Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam122c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028671.2 | NP_082947.1 | 2 - 3 | 261 | 141 | |
GenBank mRNA | AK015288.1 | - | 2 - 3 | 261 | 141 | |
AK015916.1 | - | 2 - 3 | 271 | 141 | ||
BC147640.1 | - | 2 - 3 | 249 | 9056630 | 141 | |
BC147646.1 | - | 2 - 3 | 249 | 9056636 | 141 | |
BC172114.1 | - | 2 - 3 | 249 | 9054400 | 141 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |