Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513863_g1
          See other AARSD1 GE Assays ›
          Gene Symbol:
          Aarsd1
          Gene Name:
          alanyl-tRNA synthetase domain containing 1
          Gene Aliases:
          1110069E20Rik, 2310044P18Rik, AA589600, AlaX, AlaXp-II
          Chromosome Location:
          Chr.11: 101406665 - 101417433 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513863_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aarsd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144829.1 NP_659078.1 8 - 9 908 59
          GenBank mRNA AK168281.1 - 8 - 9 875 59
          BC005711.1 - 8 - 9 908 3497962 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003804.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aarsd1

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          alanyl-tRNA aminoacylation
          regulation of translational fidelity
          tRNA aminoacylation
          nucleotide binding
          aminoacyl-tRNA editing activity
          Ser-tRNA(Ala) hydrolase activity
          nucleic acid binding
          aminoacyl-tRNA ligase activity
          alanine-tRNA ligase activity
          protein binding
          ATP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073764 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073760 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073763 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073761 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073762 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046453 AK168281.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046453 BC005711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046453 AK136332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046453 NM_144829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046453 AK009808.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49389 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133888 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173548 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046453 AK168281.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046453 BC005711.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046453 AK136332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046453 NM_144829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046453 AK009808.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073767 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133888 - -
          Whole Mouse Genome G4122A A_51_P423680 AK168281.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423680 BC005711.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423680 AK136332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P423680 NM_144829.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100615 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107730 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09310332 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09310340 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073761 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073764 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073762 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073763 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09591013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09591015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09607526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09310332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09089619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09310334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09320788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09089618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391441 - -
          Affymetrix HT MG-430 PM Array Plate 1424006_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391431 - -
          Mouse Genome 430 2.0 Array 1424006_PM_at - -
          Mouse Genome 430 2.0 Array 1424006_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69684_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69684_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100035203.mm.1 TC1100003804.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035203.mm.1 NM_144829.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035203.mm.1 NM_144829.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035204.mm.1 TC1100003804.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035204.mm.1 NM_144829.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035204.mm.1 NM_144829.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035200.mm.1 TC1100003804.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035200.mm.1 NM_144829.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035200.mm.1 NM_144829.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035202.mm.1 TC1100003804.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035202.mm.1 NM_144829.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035202.mm.1 NM_144829.1 Refseq
          Mouse Expression Set 430 1424006_at - -
          Mouse Expression Set 430 1424006_PM_at - -
          Mouse Expression Set 430 1432073_at - -
          Mouse Expression Set 430 1454256_s_at - -
          Mouse Expression Set 430 1454256_PM_s_at - -
          Mouse Expression Set 430 1432073_PM_at - -
          Murine Genome U74v2 Set 171578_PM_at - -
          Murine Genome U74v2 Set 171578_at - -
          Mouse Genome 430A 2.0 Array 1424006_PM_at - -
          Mouse Genome 430A 2.0 Array 1424006_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 653007 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230047 - -
          Illumina mouse-6 v1.0 expression beadchip 4230047 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2612702 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2612702 - -
          mouse-6 v1.1 (Illumina) scl39534.12.3_104-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39534.12.3_104-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2918440 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2918440 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39534.12.3_104-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0