Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00514153_m1
          See other PRSS44 GE Assays ›
          Gene Symbol:
          Prss44
          Gene Name:
          protease, serine 44
          Gene Aliases:
          1700036D21Rik, 79H19A, AU043574, AU043597, TESSP-4, Tessp4
          Chromosome Location:
          Chr.9: 110813900 - 110817999 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00514153_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB047758.1 - 980 95

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P11513 NM_148940.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P11513 AB162857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055112 AB047758.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825740 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967646 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149875 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114423 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P11513 NM_148940.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P11513 AB162857.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26825997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826064 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02029690 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06926188 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114423 - -
          Whole Mouse Genome G4122A A_52_P181672 NM_148940.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P181672 AB047758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P181672 AB047759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P181672 AB162857.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133898 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114423 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149875 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08119710 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07999180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07999184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26825986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07754533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07754849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08119710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07868817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07868814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07999184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26825997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826010 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10589620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589619 - -
          Affymetrix HT MG-430 PM Array Plate 1426131_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429995_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427716_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447729_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10589616 - -
          Mouse Genome 430 2.0 Array 1426131_at - -
          Mouse Genome 430 2.0 Array 1447729_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427716_PM_at - -
          Mouse Genome 430 2.0 Array 1429995_PM_at - -
          Mouse Genome 430 2.0 Array 1426131_PM_at - -
          Mouse Genome 430 2.0 Array 1447729_s_at - -
          Mouse Genome 430 2.0 Array 1429995_at - -
          Mouse Genome 430 2.0 Array 1427716_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73336_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73336_PM_at - -
          Mouse Expression Set 430 1426131_at - -
          Mouse Expression Set 430 1427716_at - -
          Mouse Expression Set 430 1427716_PM_at - -
          Mouse Expression Set 430 1426131_PM_at - -
          Mouse Expression Set 430 1447729_PM_s_at - -
          Mouse Expression Set 430 1429995_PM_at - -
          Mouse Expression Set 430 1429995_at - -
          Mouse Expression Set 430 1447729_s_at - -
          Murine Genome U74v2 Set 166931_f_at - -
          Murine Genome U74v2 Set 166931_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1426131_PM_at - -
          Mouse Genome 430A 2.0 Array 1426131_at - -
          Mouse Genome 430A 2.0 Array 1427716_PM_at - -
          Mouse Genome 430A 2.0 Array 1427716_at - -
          Affymetrix HT Mouse Genome 430A Array 1427716_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426131_at - -
          Affymetrix HT Mouse Genome 430A Array 1427716_at - -
          Affymetrix HT Mouse Genome 430A Array 1426131_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447729_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1429995_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447729_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1429995_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 508541 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060010 - -
          Illumina mouse-6 v1.0 expression beadchip 3060010 - -
          Illumina mouse-6 v1.0 expression beadchip 102060746 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242978 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254770 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242978 - -
          mouse-6 v1.1 (Illumina) scl36421.5.1_91-S - -
          Illumina Sentrix mouse-6.v1 bead array 102060746 - -
          Illumina Sentrix mouse-6.v1 bead array 3060010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36421.5.1_91-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102060746 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060010 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2837999 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2837999 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242978 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36421.5.1_91-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix