Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515198_m1
          See other IRF1 GE Assays ›
          Gene Symbol:
          Irf1
          Gene Name:
          interferon regulatory factor 1
          Gene Aliases:
          AU020929, Irf-1
          Chromosome Location:
          Chr.11: 53770014 - 53778374 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515198_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159393.1 NP_001152865.1 8 - 9 996 75
          NM_001159396.1 NP_001152868.1 8 - 9 951 75
          NM_008390.2 NP_032416.1 8 - 9 996 75
          GenBank mRNA AK152005.1 - 8 - 9 960 75
          AK152104.1 - 8 - 9 959 75
          AK152193.1 - 8 - 9 960 75
          AK153514.1 - 8 - 9 960 75
          AK155983.1 - 8 - 9 960 75
          AK157347.1 - 8 - 9 960 75
          BC003821.1 - 8 - 9 929 3600525 75
          CT010234.1 - 7 - 8 727 75
          M21065.1 - 8 - 9 932 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000614.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Irf1

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          regulation of adaptive immune response
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          apoptotic process
          I-kappaB kinase/NF-kappaB signaling
          regulation of gene expression
          positive regulation of cell death
          positive regulation of type I interferon production
          positive regulation of interferon-beta production
          positive regulation of natural killer cell differentiation
          regulation of MyD88-dependent toll-like receptor signaling pathway
          cellular response to interferon-beta
          negative regulation of tyrosine phosphorylation of STAT protein
          CD8-positive, alpha-beta T cell differentiation
          innate immune response
          regulation of innate immune response
          negative regulation of regulatory T cell differentiation
          positive regulation of T-helper 1 cell differentiation
          negative regulation of T-helper 2 cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          defense response to virus
          interferon-gamma-mediated signaling pathway
          response to growth hormone
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to peptide hormone stimulus
          regulation of CD8-positive, alpha-beta T cell proliferation
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100004893.mm.1 TC1100000614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004893.mm.1 NM_001159393.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004893.mm.1 NM_001159393.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004893.mm.1 NM_001159396.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004893.mm.1 NM_001159396.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004893.mm.1 NM_008390.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004893.mm.1 NM_008390.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004898.mm.1 TC1100000614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004898.mm.1 NM_001159393.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004898.mm.1 NM_001159393.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004894.mm.1 TC1100000614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004894.mm.1 NM_001159396.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004894.mm.1 NM_001159396.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004894.mm.1 NM_008390.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004894.mm.1 NM_008390.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004895.mm.1 TC1100000614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004895.mm.1 NM_001159396.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004895.mm.1 NM_001159396.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004895.mm.1 NM_008390.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004895.mm.1 NM_008390.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0