Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515594_mH
          See other CTSL GE Assays ›
          Gene Symbol:
          Ctsl
          Gene Name:
          cathepsin L
          Gene Aliases:
          1190035F06Rik, Ctsl1, MEP, fs, nkt
          Chromosome Location:
          Chr.13: 64363214 - 64370883 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515594_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctsl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009984.3 NP_034114.1 4 - 5 477 59
          GenBank mRNA AF121837.1 - 4 - 5 441 59
          AF121838.1 - 4 - 5 441 59
          AF121839.1 - 4 - 5 441 59
          AK002221.1 - 4 - 5 477 59
          AK011589.1 - 4 - 5 477 59
          AK049466.1 - 4 - 5 478 59
          AK136341.1 - 3 - 4 318 59
          AK145051.1 - 4 - 5 478 59
          AK147142.1 - 4 - 5 479 59
          AK150452.1 - 4 - 5 478 59
          AK151169.1 - 4 - 5 478 59
          AK153076.1 - 4 - 5 478 59
          AK159511.1 - 4 - 5 478 59
          AK160100.1 - 4 - 5 478 59
          AK161538.1 - 4 - 5 478 59
          AK165397.1 - 4 - 5 478 59
          AK169394.1 - 4 - 5 478 59
          AK169712.1 - 4 - 5 478 59
          BC068163.1 - 4 - 5 463 30359160 59
          J02583.1 - 4 - 5 454 59
          X04392.1 - 1 - 2 131 59
          X06086.1 - 4 - 5 463 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002204.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctsl

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          cell communication
          negative regulation of keratinocyte proliferation
          protein processing
          protein autoprocessing
          hair follicle morphogenesis
          catagen
          decidualization
          proteolysis involved in cellular protein catabolic process
          epithelial tube branching involved in lung morphogenesis
          regulation of actin cytoskeleton reorganization
          aminopeptidase activity
          cysteine-type endopeptidase activity
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          cysteine-type carboxypeptidase activity
          kininogen binding
          peptide binding
          histone binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300014230.mm.1 TC1300002204.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014230.mm.1 NM_009984.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014231.mm.1 TC1300002204.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014231.mm.1 NM_009984.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 908536 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix