Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517555_m1
          See other FIBP GE Assays ›
          Gene Symbol:
          Fibp
          Gene Name:
          fibroblast growth factor (acidic) intracellular binding protein
          Gene Aliases:
          2010004G08Rik, 2010005N21Rik, 3010027N18Rik
          Chromosome Location:
          Chr.19: 5460601 - 5465052 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517555_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fibp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253832.1 NP_001240761.1 3 - 4 496 75
          NM_021438.4 NP_067413.1 3 - 4 496 75
          GenBank mRNA AF270700.1 - 3 - 4 409 75
          AK008093.1 - 3 - 4 487 75
          AK012629.1 - 3 - 4 479 75
          AK028029.1 - 3 - 4 487 75
          BC031716.1 - 3 - 4 466 3969355 75
          BC049284.1 - 3 - 4 474 6395407 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000069.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fibp

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet aggregation
          fibroblast growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 BC031716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 BC049284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 AK028029.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 AF270700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 AK008132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 BC031716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 BC049284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 AK028029.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 AF270700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518246 AK008132.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47061 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162694 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 BC031716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 BC049284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 AK028029.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 AF270700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 AK008132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 BC031716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 BC049284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 AK028029.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 AF270700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518246 AK008132.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928812 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162694 - -
          Whole Mouse Genome G4122A A_51_P518246 BC031716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 BC049284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 AK028029.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 AF270700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 AK008132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 BC031716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 BC049284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 AK028029.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 AF270700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518246 AK008132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427992 BC031716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427992 BC049284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427992 AK028029.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427992 AK019399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P427992 AK012629.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125301 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120648 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162694 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14180171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14180164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14180167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14180153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14247029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14372844 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460597 - -
          Affymetrix HT MG-430 PM Array Plate 1438924_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1422516_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1438923_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430171_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460591 - -
          Mouse Genome 430 2.0 Array 1438923_PM_at - -
          Mouse Genome 430 2.0 Array 1430171_at - -
          Mouse Genome 430 2.0 Array 1422516_a_at - -
          Mouse Genome 430 2.0 Array 1438923_at - -
          Mouse Genome 430 2.0 Array 1430171_PM_at - -
          Mouse Genome 430 2.0 Array 1438924_x_at - -
          Mouse Genome 430 2.0 Array 1438924_PM_x_at - -
          Mouse Genome 430 2.0 Array 1422516_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58249_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58249_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900000830.mm.1 TC1900000069.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000830.mm.1 NM_001253832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000830.mm.1 NM_001253832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000830.mm.1 NM_021438.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000830.mm.1 NM_021438.4 Refseq
          Mouse Expression Set 430 1438923_PM_at - -
          Mouse Expression Set 430 1422516_a_at - -
          Mouse Expression Set 430 1438924_PM_x_at - -
          Mouse Expression Set 430 1438924_x_at - -
          Mouse Expression Set 430 1438923_at - -
          Mouse Expression Set 430 1422516_PM_a_at - -
          Murine Genome U74v2 Set 99176_at - -
          Murine Genome U74v2 Set 99176_PM_at - -
          Mouse Genome 430A 2.0 Array 1438923_PM_at - -
          Mouse Genome 430A 2.0 Array 1422516_a_at - -
          Mouse Genome 430A 2.0 Array 1422516_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1438923_at - -
          Mouse Genome 430A 2.0 Array 1438924_x_at - -
          Mouse Genome 430A 2.0 Array 1438924_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1422516_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1430171_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438923_at - -
          Affymetrix HT Mouse Genome 430A Array 1438923_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438924_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438924_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1422516_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1430171_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 449099 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102030338 - -
          Illumina mouse-6 v1.0 expression beadchip 102030338 - -
          Illumina mouse-6 v1.0 expression beadchip 2510450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260261 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248068 - -
          mouse-6 v1.1 (Illumina) scl52842.4.1_43-S - -
          mouse-6 v1.1 (Illumina) scl0058249.2_204-S - -
          Illumina Sentrix mouse-6.v1 bead array 2510450 - -
          Illumina Sentrix mouse-6.v1 bead array 102030338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0058249.2_204-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102030338 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2674132 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260261 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2674132 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0058249.2_204-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0