Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517751_m1
          See other DAB2 GE Assays ›
          Gene Symbol:
          Dab2
          Gene Name:
          disabled 2, mitogen-responsive phosphoprotein
          Gene Aliases:
          5730435J12Rik, AA960054, AI957090, D15Wsu122e, D630005B22Rik, Doc-2, Doc2, p96
          Chromosome Location:
          Chr.15: 6299788 - 6440715 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517751_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dab2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037905.3 NP_001032994.1 1 - 2 190 63
          NM_001102400.1 NP_001095870.1 1 - 2 190 63
          NM_023118.5 NP_075607.2 1 - 2 190 63
          GenBank mRNA AK154716.1 - 1 - 2 174 63
          AK162744.1 - 1 - 2 161 63
          AK162851.1 - 1 - 2 161 63
          AK165810.1 - 1 - 2 71 63
          AK168375.1 - 1 - 2 162 63
          BB856968.1 - 1 - 2 191 63
          CB952492.1 - 1 - 2 179 63
          U18869.1 - 1 - 2 123 63

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK154716.1, AK162744.1, AK162851.1, AK165810.1, NM_001037905.3, NM_001102400.1, NM_023118.5, U18869.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dab2

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis involved in differentiation
          in utero embryonic development
          positive regulation of receptor recycling
          positive regulation of protein phosphorylation
          transport
          endocytosis
          receptor-mediated endocytosis
          pinocytosis
          apoptotic process
          multicellular organism development
          endoderm development
          excretion
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of neuron projection development
          protein transport
          Wnt signaling pathway
          cell differentiation
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of protein binding
          positive regulation of aldosterone biosynthetic process
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of transcription elongation from RNA polymerase II promoter
          leading edge cell differentiation
          negative regulation of apoptotic process
          positive regulation of cell adhesion
          positive regulation of endocytosis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          clathrin coat assembly
          response to steroid hormone
          negative regulation of epithelial cell proliferation
          positive regulation of SMAD protein signal transduction
          negative regulation of androgen receptor signaling pathway
          negative regulation of ERK1 and ERK2 cascade
          cellular response to epidermal growth factor stimulus
          cellular response to transforming growth factor beta stimulus
          negative regulation of canonical Wnt signaling pathway
          positive regulation of substrate adhesion-dependent cell spreading
          negative regulation of protein localization to plasma membrane
          positive regulation of Wnt signaling pathway, planar cell polarity pathway
          regulation of Rho-dependent protein serine/threonine kinase activity
          positive regulation of clathrin-dependent endocytosis
          positive regulation of early endosome to late endosome transport
          positive regulation of aldosterone secretion
          positive regulation of integrin-mediated signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway
          integrin binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          protein C-terminus binding
          clathrin binding
          phosphatidylinositol binding
          AP-2 adaptor complex binding
          clathrin adaptor activity
          cargo receptor activity
          SMAD binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 TC1500000031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_001008702.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_001008702.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_001037905.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_001037905.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_001102400.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_001102400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_023118.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000235.mm.1 NM_023118.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000231.mm.1 TC1500000031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000231.mm.1 NM_001037905.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000231.mm.1 NM_001037905.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000231.mm.1 NM_001102400.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000231.mm.1 NM_001102400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000231.mm.1 NM_023118.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000231.mm.1 NM_023118.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 TC1500000031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_001008702.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_001008702.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_001037905.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_001037905.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_001102400.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_001102400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_023118.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000236.mm.1 NM_023118.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000228.mm.1 TC1500000031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000228.mm.1 NM_001037905.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000228.mm.1 NM_001037905.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000228.mm.1 NM_001102400.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000228.mm.1 NM_001102400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000228.mm.1 NM_023118.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000228.mm.1 NM_023118.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 TC1500000031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_001008702.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_001008702.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_001037905.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_001037905.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_001102400.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_001102400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_023118.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000234.mm.1 NM_023118.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 339780 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0