Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00520295_m1
          See other TXNDC5 GE Assays ›
          Gene Symbol:
          Txndc5
          Gene Name:
          thioredoxin domain containing 5
          Gene Aliases:
          AL022641, ERp46, PC-TRP
          Chromosome Location:
          Chr.13: 38500266 - 38528824 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00520295_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Txndc5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289598.1 NP_001276527.1 3 - 4 530 94
          NM_001289599.1 NP_001276528.1 3 - 4 497 94
          NM_145367.4 NP_663342.3 3 - 4 554 94
          GenBank mRNA AK136046.1 - 3 - 4 442 94
          AK142049.1 - 3 - 4 540 94
          AK150781.1 - 3 - 4 538 94
          AK169680.1 - 3 - 4 521 94
          AK224911.1 - 2 - 3 207 94
          AK224913.1 - 3 - 4 541 94
          AK224922.1 - 3 - 4 499 94
          AK224923.1 - 3 - 4 521 94
          AK224924.1 - 3 - 4 529 94
          AK224926.1 - 3 - 4 499 94
          AK224927.1 - 3 - 4 550 94
          AK224930.1 - 3 - 4 502 94
          AK224933.1 - 3 - 4 491 94
          AK224934.1 - 3 - 4 545 94
          AK224936.1 - 3 - 4 544 94
          AK224937.1 - 3 - 4 543 94
          AK224938.1 - 3 - 4 538 94
          AK224939.1 - 3 - 4 531 94
          AK224943.1 - 3 - 4 502 94
          AK224945.1 - 3 - 4 538 94
          AK224946.1 - 3 - 4 541 94
          AK224947.1 - 3 - 4 528 94
          AK224948.1 - 3 - 4 502 94
          AK224949.1 - 3 - 4 491 94
          AK224950.1 - 3 - 4 491 94
          AK224951.1 - 3 - 4 549 94
          AK224952.1 - 3 - 4 517 94
          AK224956.1 - 3 - 4 462 94
          AK224957.1 - 3 - 4 507 94
          AK224958.1 - 3 - 4 507 94
          AK224960.1 - 3 - 4 549 94
          AK224962.1 - 3 - 4 509 94
          AK224963.1 - 3 - 4 550 94
          AK224964.1 - 3 - 4 550 94
          AK224965.1 - 3 - 4 526 94
          AK224966.1 - 3 - 4 526 94
          AK224967.1 - 3 - 4 517 94
          AK224968.1 - 1 - 2 84 94
          AK224969.1 - 1 - 2 84 94
          AK224970.1 - 3 - 4 491 94
          AK224971.1 - 3 - 4 551 94
          AK224972.1 - 3 - 4 541 94
          AK224973.1 - 3 - 4 491 94
          AK224974.1 - 3 - 4 494 94
          AK224976.1 - 3 - 4 524 94
          AK224977.1 - 3 - 4 538 94
          AK224978.1 - 3 - 4 538 94
          AK224979.1 - 3 - 4 525 94
          AK224981.1 - 3 - 4 495 94
          AK224982.1 - 3 - 4 550 94
          AK224983.1 - 3 - 4 539 94
          AK224984.1 - 3 - 4 504 94
          AK224985.1 - 3 - 4 508 94
          AK224986.1 - 3 - 4 552 94
          AK224988.1 - 3 - 4 528 94
          AK224989.1 - 3 - 4 545 94
          AK224990.1 - 3 - 4 540 94
          AK224991.1 - 3 - 4 535 94
          AK224992.1 - 3 - 4 535 94
          AK224993.1 - 3 - 4 535 94
          AK224994.1 - 3 - 4 543 94
          AK224995.1 - 3 - 4 506 94
          AK224996.1 - 3 - 4 477 94
          AK224997.1 - 3 - 4 502 94
          AK224998.1 - 3 - 4 500 94
          AK224999.1 - 3 - 4 522 94
          AK225004.1 - 3 - 4 543 94
          AK225005.1 - 3 - 4 521 94
          AK225007.1 - 3 - 4 543 94
          AK225010.1 - 3 - 4 543 94
          AK225011.1 - 3 - 4 543 94
          AK225012.1 - 3 - 4 312 94
          AK225013.1 - 3 - 4 312 94
          AK225014.1 - 3 - 4 521 94
          AK225015.1 - 3 - 4 312 94
          AK225016.1 - 3 - 4 451 94
          AK225017.1 - 3 - 4 499 94
          AK225019.1 - 3 - 4 531 94
          AK225020.1 - 3 - 4 482 94
          AK225021.1 - 3 - 4 482 94
          AK225022.1 - 3 - 4 330 94
          AK225023.1 - 3 - 4 556 94
          AY243534.1 - 3 - 4 517 94
          AY548112.1 - 3 - 4 478 94
          BC016252.1 - 3 - 4 521 4911455 94
          BC024505.1 - 3 - 4 466 4983496 94
          BC036155.1 - 2 - 3 140 94
          BC046789.1 - 3 - 4 480 5716897 94
          BP765107.1 - 3 - 4 544 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001835.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Txndc5

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          response to endoplasmic reticulum stress
          apoptotic cell clearance
          cell redox homeostasis
          protein disulfide isomerase activity
          isomerase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300011450.mm.1 TC1300001835.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011450.mm.1 NM_001289598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011450.mm.1 NM_001289599.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011450.mm.1 NM_145367.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline