Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00520922_g1
          See other HAUS4 GE Assays ›
          Gene Symbol:
          Haus4
          Gene Name:
          HAUS augmin-like complex, subunit 4
          Gene Aliases:
          9430093H08Rik, AU016300, D14Ertd500e
          Chromosome Location:
          Chr.14: 54541785 - 54554361 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00520922_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Haus4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145462.2 NP_663437.2 7 - 8 915 67
          GenBank mRNA AK079161.1 - 7 - 8 855 67
          AK079852.1 - 7 - 8 883 67
          BC004644.1 - 7 - 8 890 3157901 67
          BC011095.1 - 7 - 8 890 3157901 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002058.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Haus4

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          cell cycle
          centrosome cycle
          spindle assembly
          cell division
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612962 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 AK206840.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 BC004644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 BC011095.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 NM_145462.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 AI642699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 AK206840.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 BC004644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 BC011095.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 NM_145462.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P214306 AI642699.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695946 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40423 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154321 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105867 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 AK206840.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 BC004644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 BC011095.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 NM_145462.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 AI642699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 AK206840.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 BC004644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 BC011095.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 NM_145462.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P214306 AI642699.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612975 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154321 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08116102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105867 - -
          Whole Mouse Genome G4122A A_51_P214306 AK206840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 BC004644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 BC011095.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 NM_145462.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 AI642699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 AK206840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 BC004644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 BC011095.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 NM_145462.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P214306 AI642699.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130085 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105867 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154321 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11573760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11594871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11594874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29612963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11387956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11292371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11744531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11775239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11744529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11431940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11128040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11735685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11529034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11165188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11226141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11324427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11419297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29613003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11141328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11226145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11796172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10419784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419785 - -
          Affymetrix HT MG-430 PM Array Plate 1451103_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10419779 - -
          Mouse Genome 430 2.0 Array 1451103_PM_at - -
          Mouse Genome 430 2.0 Array 1451103_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 219072_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 219072_at - -
          Affymetrix Mouse Clariom S Array PSR1400015473.mm.1 TC1400002058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015473.mm.1 NM_145462.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015473.mm.1 NM_145462.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015474.mm.1 TC1400002058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015474.mm.1 NM_145462.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015474.mm.1 NM_145462.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015475.mm.1 TC1400002058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015475.mm.1 NM_145462.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015475.mm.1 NM_145462.2 Refseq
          Mouse Expression Set 430 1451103_PM_at - -
          Mouse Expression Set 430 1451103_at - -
          Murine Genome U74v2 Set 97391_PM_at - -
          Murine Genome U74v2 Set 97391_at - -
          Mouse Genome 430A 2.0 Array 1451103_PM_at - -
          Mouse Genome 430A 2.0 Array 1451103_at - -
          Affymetrix HT Mouse Genome 430A Array 1451103_at - -
          Affymetrix HT Mouse Genome 430A Array 1451103_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2970373 - -
          Illumina mouse-6 v1.0 expression beadchip 2970373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2612044 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2612044 - -
          mouse-6 v1.1 (Illumina) scl45568.9.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45568.9.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970373 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2612044 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2612044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0