Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem161a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293797.1 | NP_001280726.1 | 4 - 5 | 372 | 61 | |
NM_145597.4 | NP_663572.1 | 5 - 6 | 476 | 61 | ||
NR_122028.1 | 5 - 6 | 476 | 61 | |||
GenBank mRNA | AK049979.1 | - | 5 - 6 | 475 | 61 | |
AK082758.1 | - | 5 - 6 | 472 | 61 | ||
AK082916.1 | - | 4 - 5 | 373 | 61 | ||
AK088569.1 | - | 4 - 5 | 362 | 61 | ||
AK147962.1 | - | 5 - 6 | 469 | 61 | ||
BC021367.1 | - | 5 - 6 | 476 | 5030434 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |