Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521874_s1
          See other MAGEB3 GE Assays ›
          Gene Symbol:
          Mageb3 Mageb1 Mageb2
          Gene Name:
          melanoma antigen, family B, 3
          melanoma antigen, family B, 1
          melanoma antigen, family B, 2
          Gene Aliases:
          Mage-b3, Mage-ps1, Mage-rs3, Smage3
          MAGE-Xp, Mage-b1, Mage-rs1, Magel1, Smage1, dam1
          Mage-b2, Mage-rs2, Smage2
          Chromosome Location:
          Chr.2: 121953771 - 121956092 on Build GRCm38
          Chromosome Location:
          Chr.X: 92015191 - 92016333 on Build GRCm38
          Chromosome Location:
          Chr.X: 91331905 - 91332897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm00521874_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mageb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008545.2 NP_032571.2 1 1325 114
          GenBank mRNA AF311315.3 - 1 593 114
          AF311316.2 - 1 593 114
          AK019658.1 - 1 1325 114
          AY099462.1 - 1 593 114
          AY099463.1 - 1 593 114
          AY099464.1 - 1 593 114
          BC141002.1 - 1 1223 9055614 114
          Gene Symbol: Mageb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010759.1 NP_034889.1 1 114
          Gene Symbol: Mageb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031171.1 NP_112448.1 1 114
          GenBank mRNA AY196960.1 - 1 114
          BC119227.1 - 1 8733976 114
          BC145851.1 - 2 40131154 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004501.mm.1, TC0X00000865.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mageb3
          Click here to find siRNAs for Gene Mageb1
          Click here to find siRNAs for Gene Mageb2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21583616 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21583618 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21583619 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172584 AY099464.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172584 AK019658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172584 AY099462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172584 AY099463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172584 NM_008545.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236677 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112421 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172584 AY099464.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172584 AK019658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172584 AY099462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172584 AY099463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172584 NM_008545.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21583594 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112421 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130630 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01879130 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01879126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21583619 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21583618 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21583616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21583617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01926765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02111643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21583591 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486897 - -
          Affymetrix HT MG-430 PM Array Plate 1450293_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605664 - -
          Affymetrix Mouse Gene 1.1 ST Array 10600726 - -
          Affymetrix Mouse Gene 1.1 ST Array 10486895 - -
          Mouse Genome 430 2.0 Array 1422345_s_at - -
          Mouse Genome 430 2.0 Array 1450293_at - -
          Mouse Genome 430 2.0 Array 1450293_PM_at - -
          Mouse Genome 430 2.0 Array 1422345_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17147_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17147_at - -
          Affymetrix Mouse Clariom S Array PSR0X00005604.mm.1 TC0X00000865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005604.mm.1 NM_031171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005604.mm.1 NM_031171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005605.mm.1 TC0X00000865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038181.mm.1 TC0200004501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038181.mm.1 NM_008545.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038181.mm.1 NM_008545.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005600.mm.1 TC0X00000865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005603.mm.1 TC0X00000865.mm.1 MTA TC
          Mouse Expression Set 430 1450293_PM_at - -
          Mouse Expression Set 430 1450293_at - -
          Mouse Expression Set 430 1422345_s_at - -
          Mouse Expression Set 430 1422345_PM_s_at - -
          Murine Genome U74v2 Set 95783_g_at - -
          Murine Genome U74v2 Set 95782_PM_at - -
          Murine Genome U74v2 Set 95782_at - -
          Murine Genome U74v2 Set 95783_PM_g_at - -
          Murine Genome U74v2 Set 95781_at - -
          Murine Genome U74v2 Set 95781_PM_at - -
          Mouse Genome 430A 2.0 Array 1422345_s_at - -
          Mouse Genome 430A 2.0 Array 1450293_PM_at - -
          Mouse Genome 430A 2.0 Array 1422345_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1450293_at - -
          Affymetrix HT Mouse Genome 430A Array 1450293_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450293_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 332181 - -
          ABI Mouse Genome Survey Microarray 516294 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2970154 - -
          MouseWG-6 v2 Expression BeadChip 70114 - -
          Illumina mouse-6 v1.0 expression beadchip 2970154 - -
          Illumina mouse-6 v1.0 expression beadchip 5570736 - -
          Illumina mouse-6 v1.0 expression beadchip 70114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2702483 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246766 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596487 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596487 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246766 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2702483 - -
          mouse-6 v1.1 (Illumina) scl0017147.2_107-S - -
          mouse-6 v1.1 (Illumina) scl017147.1_60-S - -
          mouse-6 v1.1 (Illumina) scl0017147.2_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970154 - -
          Illumina Sentrix mouse-6.v1 bead array 70114 - -
          Illumina Sentrix mouse-6.v1 bead array 5570736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017147.2_107-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl017147.1_60-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017147.2_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970154 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570736 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2702483 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2941984 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246766 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2702483 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596487 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2941984 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246766 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0