Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021921.3 | NP_068740.3 | 1 - 2 | 377 | 115 | |
GenBank mRNA | AF220195.1 | - | 1 - 2 | 96 | 115 | |
AF310135.1 | - | 1 - 2 | 158 | 115 | ||
AK014339.1 | - | 1 - 2 | 81 | 115 | ||
AK158968.1 | - | 1 - 2 | 231 | 115 | ||
BC029704.1 | - | 1 - 2 | 156 | 5359447 | 115 | |
CA317486.1 | - | 1 - 2 | 352 | 115 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |