Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00522801_m1
          See other ACSL6 GE Assays ›
          Gene Symbol:
          Acsl6
          Gene Name:
          acyl-CoA synthetase long-chain family member 6
          Gene Aliases:
          A330035H04Rik, AW050338, Facl6, LACS, Lacsl, mKIAA0837
          Chromosome Location:
          Chr.11: 54303824 - 54364756 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00522801_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acsl6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033597.1 NP_001028769.1 19 - 20 2044 66
          NM_001033598.1 NP_001028770.1 19 - 20 2013 66
          NM_001033599.1 NP_001028771.1 19 - 20 2219 66
          NM_144823.4 NP_659072.3 19 - 20 2044 66
          GenBank mRNA AB073974.1 - 18 - 19 1888 66
          AK122384.1 - 8 - 9 3810 66
          AK147283.1 - 19 - 20 2014 66
          AK147421.1 - 19 - 20 2045 66
          AY167035.1 - 19 - 20 2014 66
          AY786360.1 - 21 - 22 2562 66
          AY786361.1 - 19 - 20 1994 66
          AY786362.1 - 19 - 20 2186 66
          AY786363.1 - 19 - 20 2039 66
          BC016114.1 - 19 - 20 1970 4481949 66
          BC022959.1 - 19 - 20 2044 5362660 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000621.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acsl6

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid metabolic process
          lipid metabolic process
          fatty acid metabolic process
          triglyceride metabolic process
          neuroblast proliferation
          metabolic process
          phospholipid biosynthetic process
          positive regulation of long-chain fatty acid import across plasma membrane
          positive regulation of triglyceride biosynthetic process
          positive regulation of neuron projection development
          fatty acid transport
          nucleotide binding
          catalytic activity
          long-chain fatty acid-CoA ligase activity
          protein binding
          ATP binding
          ligase activity
          enzyme binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 TC1100000621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_001033597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_001033597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_001033598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_001033598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_001033599.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_001033599.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_144823.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004999.mm.1 NM_144823.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 TC1100000621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_001033597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_001033597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_001033598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_001033598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_001033599.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_001033599.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_144823.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005001.mm.1 NM_144823.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 TC1100000621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_001033597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_001033597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_001033598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_001033598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_001033599.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_001033599.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_144823.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004997.mm.1 NM_144823.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 TC1100000621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_001033597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_001033597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_001033598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_001033598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_001033599.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_001033599.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_144823.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005000.mm.1 NM_144823.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 TC1100000621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_001033597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_001033597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_001033598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_001033598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_001033599.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_001033599.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_144823.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004998.mm.1 NM_144823.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 TC1100000621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_001033597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_001033597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_001033598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_001033598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_001033599.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_001033599.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_144823.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005002.mm.1 NM_144823.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix