Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800020714.mm.1 |
TC0800002705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020714.mm.1 |
NM_144930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020714.mm.1 |
NM_144930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020716.mm.1 |
TC0800002705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020716.mm.1 |
NM_144930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020716.mm.1 |
NM_144930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020713.mm.1 |
TC0800002705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020713.mm.1 |
NM_144930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020713.mm.1 |
NM_144930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020717.mm.1 |
TC0800002705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020717.mm.1 |
NM_144930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020717.mm.1 |
NM_144930.2 |
Refseq |