Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800012884.mm.1 |
TC0800001569.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012884.mm.1 |
NM_145608.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012884.mm.1 |
NM_145608.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012885.mm.1 |
TC0800001569.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012885.mm.1 |
NM_145608.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012885.mm.1 |
NM_145608.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012883.mm.1 |
TC0800001569.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012883.mm.1 |
NM_145608.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012883.mm.1 |
NM_145608.2 |
Refseq |