Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525377_m1
          See other ZFP428 GE Assays ›
          Gene Symbol:
          Zfp428
          Gene Name:
          zinc finger protein 428
          Gene Aliases:
          2410005H09Rik, AV271806, AW047854, Znf428
          Chromosome Location:
          Chr.7: 24507087 - 24515682 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525377_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp428
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146183.2 NP_666295.2 2 - 3 556 114
          NR_110958.1 2 - 3 417 114
          GenBank mRNA AK010415.2 - 2 - 3 436 114
          AK139174.1 - 2 - 3 287 114
          BC024802.1 - 2 - 3 457 5360536 114
          BC096675.1 - 2 - 3 469 5146693 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000385.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp428

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005234 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005240 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005233 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005239 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005236 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005235 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005237 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093949 BC024802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093949 BC096675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093949 AK139174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093949 NM_146183.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093949 AK010415.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292238 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136781 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093949 BC024802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093949 BC096675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093949 AK139174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093949 NM_146183.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093949 AK010415.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005220 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136781 - -
          Whole Mouse Genome G4122A A_51_P181623 BC024802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P181623 BC096675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P181623 AK139174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P181623 NM_146183.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P181623 AK080517.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101177 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136781 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06187902 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06394798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06394797 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06394799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06394793 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06394800 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005240 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005234 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005233 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005235 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005236 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005237 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06394793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05972850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05972851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06394800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06103905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05998743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06103906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06465182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06394794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06573208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05981543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06394798 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550929 - -
          Affymetrix HT MG-430 PM Array Plate 1424490_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550925 - -
          Mouse Genome 430 2.0 Array 1424490_at - -
          Mouse Genome 430 2.0 Array 1424490_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232969_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232969_at - -
          Affymetrix Mouse Clariom S Array PSR0700002819.mm.1 TC0700000385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002819.mm.1 NM_146183.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002819.mm.1 NM_146183.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002821.mm.1 TC0700000385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002821.mm.1 NM_146183.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002821.mm.1 NM_146183.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002822.mm.1 TC0700000385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002822.mm.1 NM_146183.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002822.mm.1 NM_146183.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002823.mm.1 TC0700000385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002823.mm.1 NM_146183.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002823.mm.1 NM_146183.2 Refseq
          Mouse Expression Set 430 1424490_at - -
          Mouse Expression Set 430 1424490_PM_at - -
          Mouse Genome 430A 2.0 Array 1424490_PM_at - -
          Mouse Genome 430A 2.0 Array 1424490_at - -
          Affymetrix HT Mouse Genome 430A Array 1424490_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424490_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4280672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228583 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228583 - -
          mouse-6 v1.1 (Illumina) scl32960.3.40_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 4280672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32960.3.40_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228583 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2813524 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228583 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2813524 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0