Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00550281_g1
          See other MYEF2 GE Assays ›
          Gene Symbol:
          Myef2
          Gene Name:
          myelin basic protein expression factor 2, repressor
          Gene Aliases:
          9430071B01, mKIAA1341
          Chromosome Location:
          Chr.2: 125082209 - 125123661 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00550281_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myef2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162417.1 NP_001155889.1 12 - 13 1326 60
          NM_001162418.1 NP_001155890.1 11 - 12 1275 60
          NM_010852.2 NP_034982.2 11 - 12 1275 60
          GenBank mRNA AF483504.1 - 10 - 11 1001 60
          AF483505.1 - 10 - 11 1001 60
          AK012133.1 - 11 - 12 1277 60
          AK048404.1 - 12 - 13 1293 60
          AK129336.1 - 12 - 13 1238 60
          AK143230.1 - 11 - 12 1211 60
          U13262.1 - 7 - 8 716 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004521.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myef2

          Gene Ontology Categories:

          Process(es)

          RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200038417.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038417.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038417.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038417.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038417.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038417.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038417.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038431.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038431.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038431.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038431.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038431.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038431.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038431.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038421.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038421.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038421.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038421.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038421.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038421.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038421.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038435.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038435.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038435.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038435.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038435.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038435.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038435.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038418.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038418.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038418.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038418.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038418.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038418.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038418.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038433.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038433.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038433.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038433.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038433.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038433.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038433.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038422.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038422.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038422.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038422.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038422.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038422.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038422.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038424.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038424.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038424.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038424.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038424.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038424.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038424.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038426.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038426.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038426.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038426.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038426.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038426.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038426.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038419.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038419.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038419.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038425.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038425.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038425.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038425.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038425.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038425.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038425.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038430.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038430.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038430.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038430.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038430.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038430.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038430.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038423.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038423.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038423.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038423.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038423.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038423.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038423.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038428.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038428.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038428.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038428.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038428.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038428.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038428.mm.1 NM_010852.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038429.mm.1 TC0200004521.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200038429.mm.1 NM_001162417.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038429.mm.1 NM_001162417.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038429.mm.1 NM_001162418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038429.mm.1 NM_001162418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038429.mm.1 NM_010852.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200038429.mm.1 NM_010852.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 574392 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0