Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00550309_m1
          See other C4A GE Assays ›
          Gene Symbol:
          C4a C4b
          Gene Name:
          complement component 4A (Rodgers blood group)
          complement component 4B (Chido blood group)
          Gene Aliases:
          Slp
          C4, Ss
          Chromosome Location:
          Chr.17: 34809092 - 34823454 on Build GRCm38
          Chromosome Location:
          Chr.17: 34728046 - 34743901 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00550309_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011413.2 NP_035543.2 670 141
          GenBank mRNA AK129017.1 - 680 141
          M12384.1 - 4 - 5 564 141
          M21576.1 - 672 141
          Gene Symbol: C4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009780.2 NP_033910.2 5 - 6 692 141
          GenBank mRNA AK157954.1 - 5 - 6 678 141
          AK158475.1 - 5 - 6 678 141
          AK171644.1 - 5 - 6 678 141
          BC039141.1 - 5 - 6 655 141
          BC055297.1 - 5 - 6 663 141
          BC067394.1 - 5 - 6 662 30107263 141
          BC067409.1 - 5 - 6 662 30098613 141
          BC150743.1 - 5 - 6 642 9087654 141
          BC158005.1 - 5 - 6 641 9007313 141
          M11729.1 - 5 - 6 677 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002814.mm.1, TC1700002815.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C4a
          Click here to find siRNAs for Gene C4b

          Gene Ontology Categories:

          Function(s) Process(es)

          complement activation
          immunoglobulin mediated immune response
          complement binding
          complement component C1q complex binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31155963 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766392 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03310875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766445 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766490 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766399 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03310866 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766476 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03310868 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766505 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766384 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766364 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03310862 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102010 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08766353 - -
          Whole Mouse Genome G4122A A_52_P111632 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102010 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08766399 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03310862 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131724 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08766364 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08766476 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03310868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31155919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13060378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13566365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13460741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13401057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13401062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13401065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31155936 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450263 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450244 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450280 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450242 - -
          Affymetrix Mouse Clariom S Array PSR1700018022.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018022.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018022.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018016.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018016.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018016.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018019.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018019.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018019.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018024.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018024.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018024.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018085.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018085.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018085.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018086.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018086.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018086.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018087.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018087.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018087.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018023.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018023.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018023.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018017.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018017.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018017.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018018.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018018.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018018.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018089.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018089.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018089.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018092.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018092.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018092.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018015.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018015.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018015.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018020.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018020.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018020.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018021.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018021.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018021.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018090.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018090.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018090.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018091.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018091.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018091.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018014.mm.1 TC1700002814.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018014.mm.1 NM_009780.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018014.mm.1 NM_009780.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018088.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018088.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018088.mm.1 NM_011413.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018094.mm.1 TC1700002815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018094.mm.1 NM_011413.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018094.mm.1 NM_011413.2 Refseq
          Murine Genome U74v2 Set 162390_r_at - -
          Murine Genome U74v2 Set 103033_PM_at - -
          Murine Genome U74v2 Set 162390_PM_r_at - -
          Murine Genome U74v2 Set 103033_at - -
          Murine Genome U74v2 Set 137169_PM_at - -
          Murine Genome U74v2 Set 137169_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897539 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340092 - -
          Illumina mouse-6 v1.0 expression beadchip 1340092 - -
          Illumina mouse-6 v1.0 expression beadchip 5910341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340092 - -
          mouse-6 v1.1 (Illumina) scl0020567.1_162-S - -
          mouse-6 v1.1 (Illumina) scl020567.3_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910341 - -
          Illumina Sentrix mouse-6.v1 bead array 1340092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0020567.1_162-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020567.3_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340092 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2911128 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215092 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2911128 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646052 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0020567.1_162-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020567.3_14-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0