Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025686.2 | NP_079962.1 | 1 - 2 | 213 | 58 | |
GenBank mRNA | AK012680.1 | - | 1 - 2 | 214 | 58 | |
AK017767.1 | - | 1 - 2 | 216 | 58 | ||
AK028228.1 | - | 1 - 2 | 214 | 58 | ||
AK083122.1 | - | 1 - 2 | 214 | 58 | ||
AK151198.1 | - | 1 - 2 | 214 | 58 | ||
AK152842.1 | - | 1 - 2 | 233 | 58 | ||
BC021918.1 | - | 200 | 58 | |||
BC098232.1 | - | 1 - 2 | 194 | 2938123 | 58 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |