Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00551818_s1
          See other HEXIM1 GE Assays ›
          Gene Symbol:
          Hexim1
          Gene Name:
          hexamethylene bis-acetamide inducible 1
          Gene Aliases:
          7330426E13Rik, CLP-1, Clp1, HIS1
          Chromosome Location:
          Chr.11: 103116325 - 103119725 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00551818_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hexim1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138753.2 NP_620092.1 1 353 84
          GenBank mRNA AY090614.1 - 1 351 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001706.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AY090614.1, NM_138753.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hexim1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          heart development
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of signal transduction by p53 class mediator
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding
          snRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083434 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083438 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083437 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083425 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083427 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083424 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P348397 AY090614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P348397 NM_138753.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P348397 AY090614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P348397 NM_138753.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510882 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124095 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P348397 AY090614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P348397 NM_138753.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P348397 AY090614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P348397 NM_138753.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P348397 AY090614.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P348397 NM_138753.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P348397 AY090614.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P348397 NM_138753.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124095 - -
          Whole Mouse Genome G4122A A_51_P348397 AY090614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P348397 NM_138753.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P348397 AY090614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P348397 NM_138753.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85403 BC022111.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122059 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09356414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09356415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09096488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09356428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09096494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09096485 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09096496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083434 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083437 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083424 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083438 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083425 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083427 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28083430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09320879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09356423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09096485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09134689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09356425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09458818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28083445 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381699 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381701 - -
          Affymetrix HT MG-430 PM Array Plate 1425937_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1419359_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381697 - -
          Mouse Genome 430 2.0 Array 1425937_a_at - -
          Mouse Genome 430 2.0 Array 1419359_PM_at - -
          Mouse Genome 430 2.0 Array 1419359_at - -
          Mouse Genome 430 2.0 Array 1425937_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192231_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192231_at - -
          Affymetrix Mouse Clariom S Array PSR1100016122.mm.1 TC1100001706.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016122.mm.1 NM_138753.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016122.mm.1 NM_138753.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016123.mm.1 TC1100001706.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016123.mm.1 NM_138753.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016123.mm.1 NM_138753.2 Refseq
          Mouse Expression Set 430 1419359_PM_at - -
          Mouse Expression Set 430 1425937_a_at - -
          Mouse Expression Set 430 1425937_PM_a_at - -
          Mouse Expression Set 430 1419359_at - -
          Murine Genome U74v2 Set 141037_PM_at - -
          Murine Genome U74v2 Set 141037_at - -
          Mouse Genome 430A 2.0 Array 1419359_at - -
          Mouse Genome 430A 2.0 Array 1425937_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1425937_a_at - -
          Mouse Genome 430A 2.0 Array 1419359_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419359_at - -
          Affymetrix HT Mouse Genome 430A Array 1425937_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425937_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419359_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 389131 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2631994 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2631994 - -
          mouse-6 v1.1 (Illumina) scl40848.1_563-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40848.1_563-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2631994 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2631994 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40848.1_563-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0