Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024458.3 | NP_001019629.2 | 8 - 9 | 1205 | 105 | |
NM_001102444.1 | NP_001095914.1 | 8 - 9 | 1205 | 105 | ||
NM_013457.3 | NP_038485.1 | 8 - 9 | 1205 | 105 | ||
GenBank mRNA | AF096839.1 | - | 8 - 9 | 1103 | 105 | |
AF189771.1 | - | 7 - 8 | 985 | 105 | ||
AK054558.1 | - | 8 - 9 | 1057 | 105 | ||
BC034368.1 | - | 7 - 8 | 1036 | 4008924 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |