Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024458.3 | NP_001019629.2 | 9 - 10 | 1383 | 64 | |
NM_001102444.1 | NP_001095914.1 | 9 - 10 | 1383 | 64 | ||
NM_013457.3 | NP_038485.1 | 9 - 10 | 1383 | 64 | ||
GenBank mRNA | AF096839.1 | - | 9 - 10 | 1281 | 64 | |
AF189771.1 | - | 8 - 9 | 1163 | 64 | ||
AK054558.1 | - | 9 - 10 | 1235 | 64 | ||
BC034368.1 | - | 8 - 9 | 1214 | 4008924 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |