Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500010601.mm.1 |
TC1500001376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500010601.mm.1 |
NM_175134.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010601.mm.1 |
NM_175134.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010603.mm.1 |
TC1500001376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500010603.mm.1 |
NM_175134.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010603.mm.1 |
NM_175134.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010602.mm.1 |
TC1500001376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500010602.mm.1 |
NM_175134.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010602.mm.1 |
NM_175134.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010605.mm.1 |
TC1500001376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500010605.mm.1 |
NM_175134.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010605.mm.1 |
NM_175134.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010599.mm.1 |
TC1500001376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500010599.mm.1 |
NM_175134.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500010599.mm.1 |
NM_175134.4 |
Refseq |