Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003955.2 | NP_001003955.1 | 2 - 3 | 972 | 90 | |
NM_177466.4 | NP_803417.3 | 2 - 3 | 972 | 90 | ||
GenBank mRNA | AK129230.1 | - | 2 - 3 | 972 | 90 | |
AK145061.1 | - | 2 - 3 | 485 | 90 | ||
AK154922.1 | - | 2 - 3 | 955 | 90 | ||
AK218808.1 | - | 1 - 2 | 57 | 90 | ||
BC044833.1 | - | 2 - 3 | 501 | 90 | ||
BC051063.1 | - | 2 - 3 | 501 | 90 | ||
BC141380.1 | - | 2 - 3 | 902 | 90 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |