Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00626244_g1
          See other PDE4DIP GE Assays ›
          Gene Symbol:
          Pde4dip
          Gene Name:
          phosphodiesterase 4D interacting protein (myomegalin)
          Gene Aliases:
          4732458A06Rik, 9430063L05Rik, C87016, D130016K21Rik, D3Bwg1078e, Usmg4, mKIAA0454
          Chromosome Location:
          Chr.3: 97689828 - 97888707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00626244_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pde4dip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039376.2 NP_001034465.2 21 - 22 2844 76
          NM_001289701.1 NP_001276630.1 21 - 22 2844 76
          NM_001289702.1 NP_001276631.1 21 - 22 2844 76
          NM_178080.4 NP_835181.2 14 - 15 3352 76
          NR_110360.1 14 - 15 3866 76
          GenBank mRNA AK028810.1 - 14 - 15 3319 76
          AK122286.1 - 14 - 15 3347 76
          BC034266.1 - 14 - 15 2017 76
          BC050783.1 - 14 - 15 3306 76
          BC057350.1 - 14 - 15 3783 76
          BC141172.1 - 14 - 15 3280 76
          BC141185.1 - 21 - 22 2844 9055797 76
          BC145504.1 - 21 - 22 2844 9054067 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002543.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pde4dip

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein-containing complex assembly
          protein binding
          enzyme binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 TC0300002543.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 NM_001039376.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 NM_001039376.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 NM_001289701.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 NM_001289702.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 NM_178080.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 NM_178080.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019872.mm.1 NR_110360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 TC0300002543.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 NM_001039376.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 NM_001039376.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 NM_001289701.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 NM_001289702.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 NM_178080.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 NM_178080.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019869.mm.1 NR_110360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 TC0300002543.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 NM_001039376.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 NM_001039376.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 NM_001289701.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 NM_001289702.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 NM_178080.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 NM_178080.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019870.mm.1 NR_110360.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0