Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009907.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009907.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009907.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009907.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009907.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009906.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009906.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009906.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009906.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009906.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009909.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009909.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009909.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009909.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009909.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009914.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009914.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009914.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009914.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009914.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009915.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009915.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009915.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009915.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009915.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009905.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009905.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009905.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009905.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009905.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009911.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009911.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009911.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009911.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009911.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009913.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009913.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009913.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009913.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009913.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009904.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009904.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009904.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009904.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009904.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009910.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009910.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009910.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009910.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009910.mm.1 |
NM_172161.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009908.mm.1 |
TC0600001258.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009908.mm.1 |
NM_001113553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009908.mm.1 |
NM_001113553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009908.mm.1 |
NM_172161.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009908.mm.1 |
NM_172161.4 |
Refseq |