Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652182_m1
          See other GPR137B GE Assays ›
          Gene Symbol:
          Gpr137b Gpr137b-ps
          Gene Name:
          G protein-coupled receptor 137B
          G protein-coupled receptor 137B, pseudogene
          Gene Aliases:
          AI788917, AU016435, AW546472, C730041E01, C80550, C80741, D13Abb1e, Tm7sf1
          2310041G17Rik, ENSMUSG00000075118
          Chromosome Location:
          Chr.13: 13357620 - 13394236 on Build GRCm38
          Chromosome Location:
          Chr.13: 12614065 - 12650395 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00652182_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr137b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031999.2 NP_114388.2 2 - 3 547 115
          GenBank mRNA AK050369.1 - 2 - 3 488 115
          AK080884.1 - 2 - 3 500 115
          AK128965.1 - 2 - 3 547 115
          AK147921.1 - 2 - 3 566 162
          AK172286.1 - 2 - 3 492 115
          BC003212.1 - 2 - 3 540 3157256 115
          Gene Symbol: Gpr137b-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003568.1 2 - 3 526 115
          GenBank mRNA AF154337.1 - 2 - 3 510 115
          AK009736.1 - 2 - 3 512 115
          AK010724.1 - 2 - 3 525 115
          AK170103.1 - 2 - 3 520 115
          AK170650.1 - 2 - 3 428 115
          AK171335.1 - 2 - 3 520 115
          BC093499.1 - 2 - 3 507 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001451.mm.1, TC1300001461.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK147921.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr137b
          Click here to find siRNAs for Gene Gpr137b-ps

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300009259.mm.1 TC1300001451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009259.mm.1 NR_003568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009258.mm.1 TC1300001451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009258.mm.1 NR_003568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009260.mm.1 TC1300001451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009260.mm.1 NR_003568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009261.mm.1 TC1300001451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009261.mm.1 NR_003568.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009311.mm.1 TC1300001461.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009311.mm.1 NM_031999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009311.mm.1 NM_031999.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009310.mm.1 TC1300001461.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009310.mm.1 NM_031999.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009310.mm.1 NM_031999.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 373255 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0