Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00659062_m1
          See other DONSON GE Assays ›
          Gene Symbol:
          Donson
          Gene Name:
          downstream neighbor of SON
          Gene Aliases:
          1110025J21Rik, AI845729, ORF60
          Chromosome Location:
          Chr.16: 91666856 - 91688759 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00659062_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Donson
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021720.1 NP_068366.1 8 - 9 1347 130
          GenBank mRNA AF193608.1 - 5 - 6 638 130
          AK042415.1 - 7 - 8 130
          BC018792.1 - 7 - 8 1167 130
          BC043316.1 - 8 - 9 1347 5320969 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600002182.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Donson

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964441 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176585 BC018792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176585 BC043316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176585 AF193608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176585 AK042415.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176585 NM_021720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981739 AK132269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981739 BC018792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981739 BC043316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981739 AF193608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981739 AK042415.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66049 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104816 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981739 AK132269.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981739 BC018792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981739 BC043316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981739 AF193608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981739 AK042415.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176585 BC018792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176585 BC043316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176585 AF193608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176585 AK042415.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176585 NM_021720.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505169 AK132269.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505169 BC018792.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505169 BC043316.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505169 AF193608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505169 AK042415.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P285182 AK132269.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P285182 BC018792.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P285182 BC043316.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P285182 AF193608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P285182 AK042415.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30964446 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104816 - -
          Whole Mouse Genome G4122A A_52_P285182 AK132269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P285182 BC018792.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P285182 BC043316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P285182 AF193608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P285182 AK042415.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505169 AK132269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505169 BC018792.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505169 BC043316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505169 AF193608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505169 AK042415.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117945 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104816 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12871369 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12871365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12871367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12871364 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30964441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30964407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13047120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12548497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12493440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13041229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12827190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12721551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12869446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30964406 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440962 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440956 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440961 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440958 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440960 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440955 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440959 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440957 - -
          Affymetrix HT MG-430 PM Array Plate 1426739_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10440953 - -
          Mouse Genome 430 2.0 Array 1426739_at - -
          Mouse Genome 430 2.0 Array 1426739_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60364_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60364_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600016000.mm.1 TC1600002182.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016000.mm.1 NM_021720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016000.mm.1 NM_021720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016001.mm.1 TC1600002182.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600016001.mm.1 NM_021720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016001.mm.1 NM_021720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015998.mm.1 TC1600002182.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015998.mm.1 NM_021720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015998.mm.1 NM_021720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015999.mm.1 TC1600002182.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015999.mm.1 NM_021720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015999.mm.1 NM_021720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015997.mm.1 TC1600002182.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015997.mm.1 NM_021720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015997.mm.1 NM_021720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015996.mm.1 TC1600002182.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015996.mm.1 NM_021720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015996.mm.1 NM_021720.1 Refseq
          Mouse Expression Set 430 1426739_at - -
          Mouse Expression Set 430 1426739_PM_at - -
          Murine Genome U74v2 Set 166216_r_at - -
          Murine Genome U74v2 Set 166216_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1426739_at - -
          Mouse Genome 430A 2.0 Array 1426739_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426739_at - -
          Affymetrix HT Mouse Genome 430A Array 1426739_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 479947 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4780541 - -
          Illumina mouse-6 v1.0 expression beadchip 6130010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780541 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706882 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639684 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639684 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706882 - -
          mouse-6 v1.1 (Illumina) scl48203.8.22_30-S - -
          mouse-6 v1.1 (Illumina) scl0001791.1_109-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780541 - -
          Illumina Sentrix mouse-6.v1 bead array 6130010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48203.8.22_30-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001791.1_109-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130010 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780541 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2639684 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706882 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706882 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639684 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0