Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900000141.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000141.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000141.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000148.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000148.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000148.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000140.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000140.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000140.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000144.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000144.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000144.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000146.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000146.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000146.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000139.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000142.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000142.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000142.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000150.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000150.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000150.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000145.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000145.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000145.mm.1 |
NM_027545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000147.mm.1 |
TC0900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000147.mm.1 |
NM_027545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000147.mm.1 |
NM_027545.2 |
Refseq |