Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004436.mm.1 |
TC1600000468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004436.mm.1 |
NM_011749.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004436.mm.1 |
NM_011749.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004435.mm.1 |
TC1600000468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004435.mm.1 |
NM_011749.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004435.mm.1 |
NM_011749.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004433.mm.1 |
TC1600000468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004433.mm.1 |
NM_011749.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004433.mm.1 |
NM_011749.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004437.mm.1 |
TC1600000468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004437.mm.1 |
NM_011749.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004437.mm.1 |
NM_011749.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004432.mm.1 |
TC1600000468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004432.mm.1 |
NM_011749.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004432.mm.1 |
NM_011749.4 |
Refseq |