Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300015467.mm.1 |
TC1300002335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300015467.mm.1 |
NM_028493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015467.mm.1 |
NM_028493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015468.mm.1 |
TC1300002335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300015468.mm.1 |
NM_028493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015468.mm.1 |
NM_028493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015469.mm.1 |
TC1300002335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300015469.mm.1 |
NM_028493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015469.mm.1 |
NM_028493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015470.mm.1 |
TC1300002335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300015470.mm.1 |
NM_028493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015470.mm.1 |
NM_028493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015471.mm.1 |
TC1300002335.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300015471.mm.1 |
NM_028493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300015471.mm.1 |
NM_028493.2 |
Refseq |