Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00723578_m1
          See other B4GAT1 GE Assays ›
          Gene Symbol:
          B4gat1 Brms1
          Gene Name:
          beta-1,4-glucuronyltransferase 1
          breast cancer metastasis-suppressor 1
          Gene Aliases:
          1500032M01Rik, B3gnt1, B3gnt6, BETA3GNT1, iGAT, iGNT
          AV003220, AW554636
          Chromosome Location:
          Chr.19: 5038826 - 5041134 on Build GRCm38
          Chromosome Location:
          Chr.19: 5041236 - 5049917 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00723578_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B4gat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175383.2 NP_780592.1 1 - 2 1206 86
          GenBank mRNA AK050347.1 - 1 - 2 1180 86
          AK075733.1 - 1 - 2 1087 86
          AK158899.1 - 1 - 2 1207 86
          BC002191.1 - 1 - 2 534 86
          Gene Symbol: Brms1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC038380.1 - 1141 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000052.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B4gat1
          Click here to find siRNAs for Gene Brms1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          axon guidance
          protein O-linked mannosylation
          alpha-1,6-mannosyltransferase activity
          alpha-1,2-mannosyltransferase activity
          mannosyltransferase activity
          alpha-1,3-mannosyltransferase activity
          alpha-1,3-galactosyltransferase activity
          UDP-glucose:glycoprotein glucosyltransferase activity
          glycolipid mannosyltransferase activity
          oligosaccharyl transferase activity
          dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
          acetylglucosaminyltransferase activity
          acetylgalactosaminyltransferase activity
          galactosyltransferase activity
          fucosyltransferase activity
          N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase activity
          O antigen polymerase activity
          lipopolysaccharide-1,6-galactosyltransferase activity
          glucuronosyltransferase activity
          transferase activity
          glycosyltransferase activity
          cellulose synthase activity
          9-phenanthrol UDP-glucuronosyltransferase activity
          1-phenanthrol glycosyltransferase activity
          9-phenanthrol glycosyltransferase activity
          1,2-dihydroxy-phenanthrene glycosyltransferase activity
          phenanthrol glycosyltransferase activity
          beta-1,4-mannosyltransferase activity
          alpha-1,2-galactosyltransferase activity
          dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
          lipopolysaccharide-1,5-galactosyltransferase activity
          dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          inositol phosphoceramide synthase activity
          glucosyltransferase activity
          metal ion binding
          alpha-(1->3)-fucosyltransferase activity
          alpha-(1->6)-fucosyltransferase activity
          indole-3-butyrate beta-glucosyltransferase activity
          salicylic acid glucosyltransferase (ester-forming) activity
          salicylic acid glucosyltransferase (glucoside-forming) activity
          benzoic acid glucosyltransferase activity
          chondroitin hydrolase activity
          dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichol alpha-1,6-mannosyltransferase
          cytokinin 9-beta-glucosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926487 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926482 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926488 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926485 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926486 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926481 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926489 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P305320 AK050347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P305320 AK050347.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17234 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P305320 AK050347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P305320 AK050347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P305320 AK050347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P305320 AK050347.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09092300 - -
          Whole Mouse Genome G4122A A_51_P305320 AK050347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P305320 AK050347.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133418 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14193049 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14358081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14366451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14340172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14380760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14340173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14193050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14193045 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926488 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926489 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926485 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926487 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926486 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926481 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14193048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14193050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14380761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14366456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14386920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14169690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14366450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14366451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14386921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14366460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926497 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460514 - -
          Affymetrix HT MG-430 PM Array Plate 1452346_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460513 - -
          Mouse Genome 430 2.0 Array 1452346_PM_at - -
          Mouse Genome 430 2.0 Array 1452346_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108902_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108902_at - -
          Affymetrix Mouse Clariom S Array PSR1900000694.mm.1 TC1900000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000694.mm.1 NM_175383.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000694.mm.1 NM_175383.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000696.mm.1 TC1900000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000696.mm.1 NM_175383.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000696.mm.1 NM_175383.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000695.mm.1 TC1900000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000695.mm.1 NM_175383.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000695.mm.1 NM_175383.2 Refseq
          Mouse Expression Set 430 1452346_at - -
          Mouse Expression Set 430 1452346_PM_at - -
          Mouse Genome 430A 2.0 Array 1452346_PM_at - -
          Mouse Genome 430A 2.0 Array 1452346_at - -
          Affymetrix HT Mouse Genome 430A Array 1452346_at - -
          Affymetrix HT Mouse Genome 430A Array 1452346_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 912850 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257876 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257876 - -
          mouse-6 v1.1 (Illumina) scl52829.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52829.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340736 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257876 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257876 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52829.1_29-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix