Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00777368_m1
          See other CLCA3A1 GE Assays ›
          Gene Symbol:
          Clca3a1
          Gene Name:
          chloride channel calcium activated 3A1
          Gene Aliases:
          Cacc, Clca1
          Chromosome Location:
          Chr.3: 144729697 - 144760972 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00777368_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clca3a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF047838.1 - 1480 133
          AF052746.1 - 9 - 10 1509 133
          AK032511.1 - 9 - 10 1479 133
          AK087213.1 - 9 - 10 1481 133
          AK142211.1 - 9 - 10 1481 133
          AK161805.1 - 9 - 10 1482 133
          BC132342.1 - 9 - 10 1483 40130619 133
          BC145057.1 - 1483 9053605 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003251.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982291 BC010260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982291 AK142211.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982291 AK161805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982291 AK087213.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982291 AK137904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039694 AK137904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039694 AF052746.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272600 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139611 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136271 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982291 BC010260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982291 AK142211.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982291 AK161805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982291 AK087213.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982291 AK137904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A ERCC-00017_150 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P669035 BC010260.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P669035 AK142211.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P669035 AK161805.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P669035 AK087213.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P669035 AK137904.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22604930 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105043 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136271 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105096 - -
          Whole Mouse Genome G4122A A_52_P669035 BC010260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669035 AK142211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669035 AK161805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669035 AK087213.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P669035 AK137904.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114039 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132118 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05105096 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136271 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02924111 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02924121 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02924114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02924118 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02924116 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02924113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22604856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02674676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02559256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02920807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02938515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02860997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02924118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03028122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02919852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02924111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22604930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03010682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02373225 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10502553 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502558 - -
          Affymetrix HT MG-430 PM Array Plate 1417853_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417852_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10502552 - -
          Affymetrix Mouse Gene 1.1 ST Array 10502565 - -
          Mouse Genome 430 2.0 Array 1417852_PM_x_at - -
          Mouse Genome 430 2.0 Array 1437578_at - -
          Mouse Genome 430 2.0 Array 1460259_PM_s_at - -
          Mouse Genome 430 2.0 Array 1417853_at - -
          Mouse Genome 430 2.0 Array 1417853_PM_at - -
          Mouse Genome 430 2.0 Array 1417852_x_at - -
          Mouse Genome 430 2.0 Array 1437578_PM_at - -
          Mouse Genome 430 2.0 Array 1460259_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12722_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12722_at - -
          Affymetrix Mouse Clariom S Array PSR0300024172.mm.1 TC0300003251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024172.mm.1 NM_009899.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024171.mm.1 TC0300003251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024171.mm.1 NM_009899.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024176.mm.1 TC0300003251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024176.mm.1 NM_009899.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024174.mm.1 TC0300003251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024174.mm.1 NM_009899.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024173.mm.1 TC0300003251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024173.mm.1 NM_009899.4 Refseq
          Mouse Expression Set 430 1460259_s_at - -
          Mouse Expression Set 430 1460259_PM_s_at - -
          Mouse Expression Set 430 1417852_PM_x_at - -
          Mouse Expression Set 430 1417852_x_at - -
          Mouse Expression Set 430 1417853_PM_at - -
          Mouse Expression Set 430 1417853_at - -
          Murine Genome U74v2 Set 103812_at - -
          Murine Genome U74v2 Set 103812_PM_at - -
          Mouse Genome 430A 2.0 Array 1417853_at - -
          Mouse Genome 430A 2.0 Array 1417853_PM_at - -
          Mouse Genome 430A 2.0 Array 1460259_s_at - -
          Mouse Genome 430A 2.0 Array 1417852_x_at - -
          Mouse Genome 430A 2.0 Array 1417852_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1460259_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417852_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1417853_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417853_at - -
          Affymetrix HT Mouse Genome 430A Array 1417852_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1400519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731377 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731377 - -
          mouse-6 v1.1 (Illumina) scl21422.13.1_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 1400519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21422.13.1_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731377 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731377 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0