Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00780266_s1
          See other RAB5B GE Assays ›
          Gene Symbol:
          Rab5b
          Gene Name:
          RAB5B, member RAS oncogene family
          Gene Aliases:
          C030027M18Rik
          Chromosome Location:
          Chr.10: 128677172 - 128696306 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00780266_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177411.4 NP_803130.1 6 2235 95
          GenBank mRNA AK081251.1 - 6 2228 95
          AK206203.1 - 1 102 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab5b

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          endosome organization
          small GTPase mediated signal transduction
          metabolic process
          protein transport
          antigen processing and presentation
          regulation of endocytosis
          Rab protein signal transduction
          plasma membrane to endosome transport
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          GTP-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107468 BC137574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107468 BC137575.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107468 X84239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107468 AK186006.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107468 NM_177411.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084626 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407022 NM_177411.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407022 AK081251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407022 NM_177411.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407022 AK081251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870156 AK046423.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127272 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162582 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407022 NM_177411.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407022 AK081251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407022 NM_177411.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407022 AK081251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084626 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107468 BC137574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107468 BC137575.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107468 X84239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107468 AK186006.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107468 NM_177411.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542732 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07233827 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07233855 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07233818 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07233838 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07233847 - -
          Whole Mouse Genome G4122A A_52_P407022 NM_177411.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407022 AK081251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407022 NM_177411.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407022 AK081251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234263 BC137574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234263 BC137575.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234263 BY727964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234263 X84239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234263 AK083034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P92347 AK083034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P92347 AK144551.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07233855 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07233838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08819366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08839749 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373525 - -
          Affymetrix HT MG-430 PM Array Plate 1422119_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437446_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10486057 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373521 - -
          Mouse Genome 430 2.0 Array 1443223_PM_at - -
          Mouse Genome 430 2.0 Array 1422119_PM_at - -
          Mouse Genome 430 2.0 Array 1437446_at - -
          Mouse Genome 430 2.0 Array 1422119_at - -
          Mouse Genome 430 2.0 Array 1443223_at - -
          Mouse Genome 430 2.0 Array 1437446_PM_at - -
          Mouse Expression Set 430 1422119_PM_at - -
          Mouse Expression Set 430 1422119_at - -
          Mouse Expression Set 430 1437446_at - -
          Mouse Expression Set 430 1437446_PM_at - -
          Murine Genome U74v2 Set 98731_PM_at - -
          Murine Genome U74v2 Set 98731_at - -
          Mouse Genome 430A 2.0 Array 1422119_at - -
          Mouse Genome 430A 2.0 Array 1422119_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422119_at - -
          Affymetrix HT Mouse Genome 430A Array 1422119_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437446_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437446_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 365772 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6900731 - -
          Illumina mouse-6 v1.0 expression beadchip 6900731 - -
          Illumina mouse-6 v1.0 expression beadchip 1980273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006900731 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163335 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218559 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604256 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218559 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604256 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3163335 - -
          mouse-6 v1.1 (Illumina) scl37351.6_4-S - -
          mouse-6 v1.1 (Illumina) scl0003920.1_1369-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980273 - -
          Illumina Sentrix mouse-6.v1 bead array 6900731 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003920.1_1369-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37351.6_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980273 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900731 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3163335 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604256 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218559 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163335 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003920.1_1369-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37351.6_4-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0