Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00788073_s1
          See other YAE1D1 GE Assays ›
          Gene Symbol:
          Yae1d1
          Gene Name:
          Yae1 domain containing 1
          Gene Aliases:
          1600012F09Rik
          Chromosome Location:
          Chr.13: 17986640 - 17993351 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00788073_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Yae1d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025904.3 NP_080180.1 3 1823 84
          GenBank mRNA AK005425.1 - 3 1811 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001509.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Yae1d1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854602 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854603 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854600 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854604 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963634 NM_025904.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963634 AK005425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963631 AK151981.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963631 AK216868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963631 AK076034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963631 BC048453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963631 AK011852.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525638 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963631 AK151981.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963631 AK216868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963631 AK076034.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963631 BC048453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963631 AK011852.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963634 NM_025904.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963634 AK005425.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854619 - -
          Whole Mouse Genome G4122A A_52_P529098 AK030455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309618 NM_025904.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P309618 AK005425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P269956 AK151981.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P269956 AK076034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P269956 BC048453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P269956 AK011852.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P269956 AK019184.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11106205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11106212 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11106211 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11106204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11106208 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854602 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854600 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854603 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854604 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28854599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28854580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10453421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10453416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10453419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10693353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10782337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10820896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28854583 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403760 - -
          Affymetrix HT MG-430 PM Array Plate 1419660_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419661_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449557_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10407916 - -
          Affymetrix Mouse Gene 1.1 ST Array 10497587 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403756 - -
          Mouse Genome 430 2.0 Array 1449557_at - -
          Mouse Genome 430 2.0 Array 1419661_at - -
          Mouse Genome 430 2.0 Array 1419660_PM_at - -
          Mouse Genome 430 2.0 Array 1449557_PM_at - -
          Mouse Genome 430 2.0 Array 1419660_at - -
          Mouse Genome 430 2.0 Array 1419661_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300009564.mm.1 TC1300001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009564.mm.1 NM_025904.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009564.mm.1 NM_025904.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009566.mm.1 TC1300001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009566.mm.1 NM_025904.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009566.mm.1 NM_025904.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009562.mm.1 TC1300001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009562.mm.1 NM_025904.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009562.mm.1 NM_025904.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009568.mm.1 TC1300001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009568.mm.1 NM_025904.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009568.mm.1 NM_025904.3 Refseq
          Mouse Expression Set 430 1419661_at - -
          Mouse Expression Set 430 1419661_PM_at - -
          Mouse Expression Set 430 1449557_PM_at - -
          Mouse Expression Set 430 1419660_PM_at - -
          Mouse Expression Set 430 1419660_at - -
          Mouse Expression Set 430 1449557_at - -
          Murine Genome U74v2 Set 168861_PM_at - -
          Murine Genome U74v2 Set 168861_at - -
          Mouse Genome 430A 2.0 Array 1419661_at - -
          Mouse Genome 430A 2.0 Array 1449557_PM_at - -
          Mouse Genome 430A 2.0 Array 1419660_PM_at - -
          Mouse Genome 430A 2.0 Array 1419661_PM_at - -
          Mouse Genome 430A 2.0 Array 1419660_at - -
          Mouse Genome 430A 2.0 Array 1449557_at - -
          Affymetrix HT Mouse Genome 430A Array 1449557_at - -
          Affymetrix HT Mouse Genome 430A Array 1419660_at - -
          Affymetrix HT Mouse Genome 430A Array 1419661_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419660_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419661_at - -
          Affymetrix HT Mouse Genome 430A Array 1449557_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 860708 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4120463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2683147 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2683147 - -
          mouse-6 v1.1 (Illumina) scl067008.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 4120463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067008.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120463 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2683155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix