Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00801707_m1
          See other EMP2 GE Assays ›
          Gene Symbol:
          Emp2
          Gene Name:
          epithelial membrane protein 2
          Gene Aliases:
          XMP
          Chromosome Location:
          Chr.16: 10281749 - 10313968 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00801707_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007929.2 NP_031955.2 1 - 2 154 63
          GenBank mRNA AK076194.1 - 1 - 2 133 63
          AK077260.1 - 1 - 2 156 63
          BC041670.1 - 1 - 2 57 3663158 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001191.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK076194.1, AK077260.1, BC041670.1, NM_007929.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emp2

          Gene Ontology Categories:

          Function(s) Process(es)

          membrane raft assembly
          T cell mediated cytotoxicity
          regulation of cell-matrix adhesion
          positive regulation of cell-matrix adhesion
          regulation of glomerular filtration
          actin filament organization
          cell adhesion
          cell-matrix adhesion
          embryo implantation
          cell death
          positive regulation of cell population proliferation
          regulation of endothelial cell migration
          cell migration
          bleb assembly
          activation of protein kinase activity
          protein localization to cell surface
          blood vessel endothelial cell migration
          regulation of kinase activity
          early endosome to late endosome transport
          regulation of angiogenesis
          actin-mediated cell contraction
          caveola assembly
          protein localization to plasma membrane
          positive regulation of integrin-mediated signaling pathway
          regulation of vasculogenesis
          integrin binding
          kinase binding
          protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600009478.mm.1 TC1600001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009478.mm.1 NM_007929.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009478.mm.1 NM_007929.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009476.mm.1 TC1600001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009476.mm.1 NM_007929.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009476.mm.1 NM_007929.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009477.mm.1 TC1600001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009477.mm.1 NM_007929.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009477.mm.1 NM_007929.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0