Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016601.mm.1 |
TC1500002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016601.mm.1 |
NM_009713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016601.mm.1 |
NM_009713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016599.mm.1 |
TC1500002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016599.mm.1 |
NM_009713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016599.mm.1 |
NM_009713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016605.mm.1 |
TC1500002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016605.mm.1 |
NM_009713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016605.mm.1 |
NM_009713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016604.mm.1 |
TC1500002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016604.mm.1 |
NM_009713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016604.mm.1 |
NM_009713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016598.mm.1 |
TC1500002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016598.mm.1 |
NM_009713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016598.mm.1 |
NM_009713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016602.mm.1 |
TC1500002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016602.mm.1 |
NM_009713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016602.mm.1 |
NM_009713.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016603.mm.1 |
TC1500002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016603.mm.1 |
NM_009713.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016603.mm.1 |
NM_009713.4 |
Refseq |