Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00802543_g1
          See other EPHB3 GE Assays ›
          Gene Symbol:
          Ephb3
          Gene Name:
          Eph receptor B3
          Gene Aliases:
          AW456895, Cek10, Etk2, HEK2, MDK5, Sek4, Tyro6
          Chromosome Location:
          Chr.16: 21204795 - 21223309 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00802543_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ephb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010143.1 NP_034273.1 14 - 15 3091 88
          GenBank mRNA AK144414.1 - 14 - 15 3108 88
          AK158043.1 - 15 - 16 4030 88
          BC014822.1 - 14 - 15 3092 3673003 88
          BC027805.1 - 10 - 11 2378 88
          BC053085.1 - 14 - 15 3091 5708827 88
          U11493.1 - 12 - 13 2551 88
          X76012.1 - 3 - 4 562 88
          Z49086.1 - 14 - 15 2949 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000298.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ephb3

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          urogenital system development
          protein phosphorylation
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          multicellular organism development
          nervous system development
          axon guidance
          axonal fasciculation
          phosphorylation
          cell migration
          peptidyl-tyrosine phosphorylation
          central nervous system projection neuron axonogenesis
          corpus callosum development
          regulation of cell-cell adhesion
          retinal ganglion cell axon guidance
          substrate adhesion-dependent cell spreading
          regulation of GTPase activity
          protein autophosphorylation
          ephrin receptor signaling pathway
          thymus development
          digestive tract morphogenesis
          regulation of axonogenesis
          positive regulation of synapse assembly
          roof of mouth development
          dendritic spine development
          dendritic spine morphogenesis
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          ephrin receptor activity
          transmembrane-ephrin receptor activity
          ATP binding
          axon guidance receptor activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600002971.mm.1 TC1600000298.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600002971.mm.1 NM_010143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002971.mm.1 NM_010143.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix