Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00802639_m1
          See other RABAC1 GE Assays ›
          Gene Symbol:
          Rabac1
          Gene Name:
          Rab acceptor 1 (prenylated)
          Gene Aliases:
          2310040I06Rik, Gbpap1, PRA1, prenylin
          Chromosome Location:
          Chr.7: 24969750 - 24972728 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00802639_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rabac1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010261.2 NP_034391.1 3 - 4 509 69
          GenBank mRNA AF120162.1 - 3 - 4 453 69
          AF252856.1 - 3 - 4 443 69
          AK008559.1 - 3 - 4 421 69
          AK009726.1 - 3 - 4 369 69
          AK046109.1 - 2 - 3 405 69
          AK150681.1 - 3 - 4 390 69
          AK170289.1 - 3 - 4 469 69
          BC008242.1 - 3 - 4 442 2655184 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002615.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rabac1

          Gene Ontology Categories:

          Process(es)

          protein binding
          protein C-terminus binding
          identical protein binding
          proline-rich region binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007550 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007636 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007547 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007548 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007552 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007551 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955227 AK009726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955227 AK046109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955227 AK150681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955227 AK170289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955227 BC008242.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043723 AK009726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043723 AK046109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043723 AK150681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043723 AK170289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043723 BC008242.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80368 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155406 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043723 AK009726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043723 AK046109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043723 AK150681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043723 AK170289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043723 BC008242.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955227 AK009726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955227 AK046109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955227 AK150681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955227 AK170289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955227 BC008242.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007662 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155406 - -
          Whole Mouse Genome G4122A A_51_P374383 AF252856.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111099 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155406 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06098578 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06344670 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06098563 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06344668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06098579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007550 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007547 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007548 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007552 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007551 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007636 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25007633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06077040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05955752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06098579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06098568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05985980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06344670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06098572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06076709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06076706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06098567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05955756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06098566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007656 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560913 - -
          Affymetrix HT MG-430 PM Array Plate 1427773_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560911 - -
          Mouse Genome 430 2.0 Array 1427773_PM_a_at - -
          Mouse Genome 430 2.0 Array 1427773_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14470_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14470_at - -
          Affymetrix Mouse Clariom S Array PSR0700022548.mm.1 TC0700002615.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022548.mm.1 NM_010261.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022548.mm.1 NM_010261.2 Refseq
          Mouse Expression Set 430 1427773_PM_a_at - -
          Mouse Expression Set 430 1427773_a_at - -
          Murine Genome U74v2 Set 94269_at - -
          Murine Genome U74v2 Set 94269_PM_at - -
          Murine Genome U74v2 Set 168504_r_at - -
          Murine Genome U74v2 Set 167883_PM_at - -
          Murine Genome U74v2 Set 168504_PM_r_at - -
          Murine Genome U74v2 Set 167883_at - -
          Mouse Genome 430A 2.0 Array 1427773_a_at - -
          Mouse Genome 430A 2.0 Array 1427773_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427773_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427773_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913030 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2100632 - -
          MouseWG-6 v2 Expression BeadChip 2030435 - -
          Illumina mouse-6 v1.0 expression beadchip 2030435 - -
          Illumina mouse-6 v1.0 expression beadchip 2100632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002100632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244857 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710113 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244857 - -
          mouse-6 v1.1 (Illumina) scl00015.1_21-S - -
          mouse-6 v1.1 (Illumina) scl014470.4_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 2100632 - -
          Illumina Sentrix mouse-6.v1 bead array 2030435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014470.4_28-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00015.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2100632 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244857 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710112 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710112 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244857 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl014470.4_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00015.1_21-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0