Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271397.1 | NP_001258326.1 | 9 - 10 | 3159 | 82 | |
NR_073167.1 | 9 - 10 | 3340 | 82 | |||
NR_073168.1 | 9 - 10 | 3155 | 82 | |||
GenBank mRNA | AK017551.1 | - | 2 - 3 | 236 | 82 | |
AK029961.1 | - | 9 - 10 | 2872 | 82 | ||
AK147172.1 | - | 9 - 10 | 2895 | 82 | ||
BC049166.1 | - | 9 - 10 | 2749 | 82 | ||
BC058699.1 | - | 9 - 10 | 3315 | 82 | ||
BC125575.1 | - | 9 - 10 | 2885 | 40130090 | 82 | |
BC125579.1 | - | 9 - 10 | 2885 | 40130094 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |