Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805335_g1
          See other FGB GE Assays ›
          Gene Symbol:
          Fgb
          Gene Name:
          fibrinogen beta chain
          Gene Aliases:
          2510049G14Rik
          Chromosome Location:
          Chr.3: 83042303 - 83049790 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805335_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181849.2 NP_862897.1 2 - 3 307 66
          GenBank mRNA AK011118.1 - 2 - 3 327 66
          AK011920.1 - 2 - 3 323 66
          AK146960.1 - 2 - 3 327 66
          AK168626.1 - 2 - 3 321 66
          BC031715.1 - 2 - 3 307 4195310 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002230.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgb

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          cell-matrix adhesion
          signal transduction
          blood coagulation
          hemostasis
          platelet activation
          positive regulation of heterotypic cell-cell adhesion
          cellular protein-containing complex assembly
          induction of bacterial agglutination
          innate immune response
          positive regulation of vasoconstriction
          positive regulation of exocytosis
          positive regulation of protein secretion
          protein polymerization
          response to calcium ion
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          positive regulation of peptide hormone secretion
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell apoptotic process
          signaling receptor binding
          structural molecule activity
          protein-macromolecule adaptor activity
          cell adhesion molecule binding
          chaperone binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300016763.mm.1 TC0300002230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016763.mm.1 NM_181849.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016762.mm.1 TC0300002230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016762.mm.1 NM_181849.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016764.mm.1 TC0300002230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016764.mm.1 NM_181849.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932273 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0