Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009768.2 | NP_033898.1 | 2 - 3 | 581 | 85 | |
GenBank mRNA | AY089967.1 | - | 2 - 3 | 439 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |