Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00840968_g1
          See other FLCN GE Assays ›
          Gene Symbol:
          Flcn
          Gene Name:
          folliculin
          Gene Aliases:
          AU014660, B430214A04Rik, Bhd, FLCL
          Chromosome Location:
          Chr.11: 59791408 - 59810039 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00840968_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Flcn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271356.1 NP_001258285.1 11 - 12 1908 86
          NM_001271357.1 NP_001258286.1 11 - 12 1651 86
          NM_146018.2 NP_666130.1 11 - 12 1948 86
          NR_073164.1 11 - 12 2219 86
          GenBank mRNA AK039106.1 - 11 - 12 1735 86
          AK080933.1 - 11 - 12 1733 86
          AK136071.1 - 11 - 12 1639 86
          AK148216.1 - 11 - 12 1702 86
          AK154040.1 - 11 - 12 1930 86
          AK161460.1 - 11 - 12 1991 86
          AK172245.1 - 11 - 12 1706 86
          BC025820.1 - 11 - 12 1682 5099493 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002862.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Flcn

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          in utero embryonic development
          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          cell-cell junction assembly
          negative regulation of gene expression
          negative regulation of mitochondrion organization
          negative regulation of angiogenesis
          hemopoiesis
          negative regulation of cell growth
          negative regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          TOR signaling
          regulation of TOR signaling
          negative regulation of TOR signaling
          positive regulation of TOR signaling
          regulation of cytokinesis
          negative regulation of Rho protein signal transduction
          regulation of histone acetylation
          positive regulation of cell adhesion
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          negative regulation of protein kinase B signaling
          negative regulation of ERK1 and ERK2 cascade
          energy homeostasis
          negative regulation of protein localization to nucleus
          negative regulation of cell proliferation involved in kidney development
          negative regulation of cellular respiration
          negative regulation of mitochondrial DNA metabolic process
          negative regulation of muscle tissue development
          negative regulation of post-translational protein modification
          regulation of pro-B cell differentiation
          negative regulation of ATP biosynthetic process
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100025942.mm.1 TC1100002862.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025942.mm.1 NM_001271356.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025942.mm.1 NM_001271357.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025942.mm.1 NM_146018.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025942.mm.1 NR_073164.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0