Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00969586_m1
          See other CNOT2 GE Assays ›
          Gene Symbol:
          Cnot2
          Gene Name:
          CCR4-NOT transcription complex, subunit 2
          Gene Aliases:
          2600016M12Rik, 2810470K03Rik, AA537049, AA959607, AW557563, C79650
          Chromosome Location:
          Chr.10: 116485161 - 116581535 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00969586_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnot2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037846.3 NP_001032935.2 5 - 6 539 89
          NM_001037847.2 NP_001032936.2 5 - 6 469 89
          NM_028082.2 NP_082358.2 4 - 5 416 89
          NR_024327.1 3 - 4 349 89
          GenBank mRNA AK011231.1 - 3 - 4 339 89
          AK078121.1 - 5 - 6 529 89
          AK167834.1 - 5 - 6 517 89
          BC043133.1 - 5 - 6 440 89
          BC063105.1 - 6 - 7 682 5003026 89
          BC065171.1 - 5 - 6 469 6415034 89
          BC090624.1 - 3 - 4 345 6815664 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002907.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          trophectodermal cell differentiation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of translation
          multicellular organism development
          positive regulation of cytoplasmic mRNA processing body assembly
          gene silencing by RNA
          negative regulation of intracellular estrogen receptor signaling pathway
          RNA phosphodiester bond hydrolysis, exonucleolytic
          regulation of stem cell population maintenance
          transcription corepressor binding
          poly(A)-specific ribonuclease activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 TC1000002907.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NM_001037846.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NM_001037846.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NM_001037847.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NM_001037847.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NM_028082.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NM_028082.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NR_024327.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021980.mm.1 NR_024327.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 TC1000002907.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NM_001037846.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NM_001037846.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NM_001037847.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NM_001037847.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NM_028082.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NM_028082.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NR_024327.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021979.mm.1 NR_024327.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0