Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252266.1 | NP_001239195.1 | 4 - 5 | 458 | 65 | |
NM_010492.3 | NP_034622.3 | 5 - 6 | 871 | 65 | ||
GenBank mRNA | AB303567.1 | - | 4 - 5 | 382 | 65 | |
AK005965.1 | - | 5 - 6 | 576 | 65 | ||
AK016989.1 | - | 5 - 6 | 574 | 65 | ||
AK206264.1 | - | 3 - 4 | 251 | 65 | ||
BC002030.1 | - | 5 - 6 | 505 | 3487814 | 65 | |
DV659030.1 | - | 3 - 4 | 243 | 65 | ||
U26459.1 | - | 5 - 6 | 494 | 65 | ||
U26460.1 | - | 4 - 5 | 400 | 65 | ||
U26461.1 | - | 4 - 5 | 399 | 65 | ||
U34595.1 | - | 3 - 4 | 367 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |