Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01136685_g1
          See other ARFIP2 GE Assays ›
          Gene Symbol:
          Arfip2
          Gene Name:
          ADP-ribosylation factor interacting protein 2
          Gene Aliases:
          2310002N04Rik, POR, Por1
          Chromosome Location:
          Chr.7: 105634201 - 105640416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01136685_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arfip2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029802.4 NP_084078.3 7 - 8 1085 82
          GenBank mRNA AK009100.1 - 7 - 8 1023 82
          AK154772.1 - 7 - 8 1051 82
          AK182646.1 - 87 83
          BC013794.1 - 7 - 8 996 2648132 82
          BC022942.1 - 7 - 8 1009 3583149 82
          BC028310.1 - 7 - 8 1000 82
          BC034520.1 - 7 - 8 1066 3964827 82
          BC079603.1 - 4 - 5 2258 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003985.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arfip2

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton organization
          ruffle organization
          regulation of Arp2/3 complex-mediated actin nucleation
          protein binding
          protein domain specific binding
          GTP-dependent protein binding
          phosphatidylinositol-4-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25393462 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25393463 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25393461 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066234 BC022942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066234 BC028310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066234 BC034520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066234 BC079603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066234 AK009100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140423 BC022942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140423 BC028310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140423 BC034520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140423 BC079603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140423 AK009100.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152871 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111197 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066234 BC022942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066234 BC028310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066234 BC034520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066234 BC079603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066234 AK009100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140423 BC022942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140423 BC028310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140423 BC034520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140423 BC079603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140423 AK009100.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25393451 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111197 - -
          Whole Mouse Genome G4122A A_51_P326425 BC022942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326425 BC028310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326425 BC034520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326425 BC079603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326425 AK009100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29785 BC022942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29785 BC028310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29785 BC034520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29785 BC079603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29785 AK009100.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135848 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123829 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06392895 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06009222 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06392901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06392898 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06009214 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06009200 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06009219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25393461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25393462 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25393463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25393408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06392906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06392889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06271765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06271761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06009219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25393424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06271740 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566504 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566507 - -
          Affymetrix HT MG-430 PM Array Plate 1435498_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424240_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10566502 - -
          Mouse Genome 430 2.0 Array 1435498_PM_at - -
          Mouse Genome 430 2.0 Array 1435498_at - -
          Mouse Genome 430 2.0 Array 1424240_PM_at - -
          Mouse Genome 430 2.0 Array 1424240_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76932_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76932_at - -
          Affymetrix Mouse Clariom S Array PSR0700033197.mm.1 TC0700003985.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033197.mm.1 NM_029802.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033197.mm.1 NM_029802.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033195.mm.1 TC0700003985.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033195.mm.1 NM_029802.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033195.mm.1 NM_029802.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033199.mm.1 TC0700003985.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033199.mm.1 NM_029802.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033199.mm.1 NM_029802.4 Refseq
          Mouse Expression Set 430 1424240_at - -
          Mouse Expression Set 430 1424240_PM_at - -
          Mouse Expression Set 430 1435498_at - -
          Mouse Expression Set 430 1435498_PM_at - -
          Murine Genome U74v2 Set 99497_PM_at - -
          Murine Genome U74v2 Set 99497_at - -
          Murine Genome U74v2 Set 166921_f_at - -
          Murine Genome U74v2 Set 166921_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1424240_at - -
          Mouse Genome 430A 2.0 Array 1424240_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424240_at - -
          Affymetrix HT Mouse Genome 430A Array 1424240_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435498_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435498_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1850301 - -
          Illumina mouse-6 v1.0 expression beadchip 4670180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850301 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619491 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233495 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233495 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2619491 - -
          mouse-6 v1.1 (Illumina) scl30883.9.1_56-S - -
          mouse-6 v1.1 (Illumina) scl00013.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670180 - -
          Illumina Sentrix mouse-6.v1 bead array 1850301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00013.1_82-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30883.9.1_56-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670180 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233495 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2955806 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2619491 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2955806 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233495 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2619491 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30883.9.1_56-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0