Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01137297_m1
          See other UMAD1 GE Assays ›
          Gene Symbol:
          Umad1
          Gene Name:
          UMAP1-MVP12 associated (UMA) domain containing 1
          Gene Aliases:
          Gm16039, OTTMUSG00000029336
          Chromosome Location:
          Chr.6: 8259288 - 8428767 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01137297_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Umad1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK158918.1 - 366 88
          BY291850.1 - 3 - 4 375 88

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 BC061003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 AK136084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 NR_033518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 AK009885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 BC061003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 AK136084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 NR_033518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P62011 AK009885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 BC061003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 AK136084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 NR_033518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 AK009885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 BC061003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 AK136084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 NR_033518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62011 AK009885.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05215219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05015579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05825387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05522284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05711113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05245705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05487126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05825391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05551213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05849070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05215215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05378307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05088778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05088792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05761195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05798455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05107485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05711121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05793425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05215218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05015580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05458921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05088779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05798449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05460374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05589437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05551214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05204426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05088798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05551212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05487127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05849072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05460372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05267867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05159379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05107506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05088790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05282317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05406504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05107487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05074571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05047419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05865269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05682381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24226010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24225247 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10536394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536392 - -
          Affymetrix HT MG-430 PM Array Plate 1428603_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443555_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457061_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10536390 - -
          Mouse Genome 430 2.0 Array 1457061_PM_at - -
          Mouse Genome 430 2.0 Array 1457061_at - -
          Mouse Genome 430 2.0 Array 1428603_PM_at - -
          Mouse Genome 430 2.0 Array 1428603_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100036521_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100036521_PM_at - -
          Mouse Expression Set 430 1457061_PM_at - -
          Mouse Expression Set 430 1428603_PM_at - -
          Mouse Expression Set 430 1428603_at - -
          Mouse Expression Set 430 1443555_PM_at - -
          Mouse Expression Set 430 1443555_at - -
          Mouse Expression Set 430 1457061_at - -
          Affymetrix HT Mouse Genome 430B Array 1457061_at - -
          Affymetrix HT Mouse Genome 430B Array 1428603_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457061_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428603_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101690148 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260397 - -
          Illumina Sentrix mouse-6.v1 bead array 101690148 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101690148 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260397 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0