Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01138208_g1
          See other HIRIP3 GE Assays ›
          Gene Symbol:
          Hirip3
          Gene Name:
          HIRA interacting protein 3
          Gene Aliases:
          B130036O03, C86302
          Chromosome Location:
          Chr.7: 126861972 - 126865377 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01138208_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hirip3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172746.3 NP_766334.1 6 - 7 2117 67
          GenBank mRNA AK045122.1 - 6 - 7 1652 67
          AK135526.1 - 6 - 7 1662 67
          AK161657.1 - 6 - 7 1652 67
          AK162096.1 - 3 - 4 679 67
          BC055687.1 - 6 - 7 2117 6414655 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001822.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hirip3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502650 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502656 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 BC055687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 AK161657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 AK162096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 AK135526.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 NM_172746.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 BC055687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 AK161657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 AK162096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 AK135526.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165683 NM_172746.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491984 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159863 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 BC055687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 AK161657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 AK162096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 AK135526.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 NM_172746.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 BC055687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 AK161657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 AK162096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 AK135526.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165683 NM_172746.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159863 - -
          Whole Mouse Genome G4122A A_51_P165683 BC055687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 AK161657.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 AK162096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 AK135526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 NM_172746.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 BC055687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 AK161657.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 AK162096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 AK135526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165683 NM_172746.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131207 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159863 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06333481 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06333483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06333484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06333499 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06333477 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06333474 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502650 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06655202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06655200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05909597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06333479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06298032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06333501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06188039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06553073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06553061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06656830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06192460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06018785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05909593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06192452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557521 - -
          Affymetrix HT MG-430 PM Array Plate 1434936_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10557519 - -
          Mouse Genome 430 2.0 Array 1434936_PM_at - -
          Mouse Genome 430 2.0 Array 1434936_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233876_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233876_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700015612.mm.1 TC0700001822.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015612.mm.1 NM_172746.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015612.mm.1 NM_172746.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015613.mm.1 TC0700001822.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015613.mm.1 NM_172746.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015613.mm.1 NM_172746.3 Refseq
          Mouse Expression Set 430 1434936_at - -
          Mouse Expression Set 430 1434936_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434936_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434936_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 782101 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 940066 - -
          Illumina mouse-6 v1.0 expression beadchip 940066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215898 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215898 - -
          mouse-6 v1.1 (Illumina) scl32046.5.1_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 940066 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32046.5.1_83-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215898 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215898 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32046.5.1_83-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline