Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01149000_g1
          See other LSM7 GE Assays ›
          Gene Symbol:
          Lsm7
          Gene Name:
          LSM7 homolog, U6 small nuclear RNA associated (S. cerevisiae)
          Gene Aliases:
          0910001B06Rik, 1110033F18Rik
          Chromosome Location:
          Chr.10: 80852823 - 80855249 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01149000_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsm7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025349.2 NP_079625.1 3 - 4 195 61
          GenBank mRNA AK003062.1 - 3 - 4 183 61
          AK007462.1 - 3 - 4 192 61
          AK009317.1 - 3 - 4 182 61
          AK012446.1 - 3 - 4 192 61
          AK013355.1 - 3 - 4 196 61
          BC027511.1 - 3 - 4 185 1382565 61
          BC081444.1 - 2 - 3 178 5711629 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002517.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsm7

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA catabolic process
          protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285906 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285904 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003443 AK007462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003443 AK009317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003443 AK003062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003443 AK012446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003443 BC027511.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65726 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156871 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003443 AK007462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003443 AK009317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003443 AK003062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003443 AK012446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003443 BC027511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P200044 AK013355.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285894 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156871 - -
          Whole Mouse Genome G4122A A_51_P200044 AK013355.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113493 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131378 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08373885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08373895 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08373889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08373888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285906 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285891 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371005 - -
          Affymetrix HT MG-430 PM Array Plate 1417313_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371002 - -
          Affymetrix Mouse Gene 1.1 ST Array 10364373 - -
          Mouse Genome 430 2.0 Array 1417313_PM_at - -
          Mouse Genome 430 2.0 Array 1417313_at - -
          Affymetrix Mouse Clariom S Array PSR1000018777.mm.1 TC1000002517.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018777.mm.1 NM_025349.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018777.mm.1 NM_025349.2 Refseq
          Mouse Expression Set 430 1417313_PM_at - -
          Mouse Expression Set 430 1417313_at - -
          Murine Genome U74v2 Set 97907_at - -
          Murine Genome U74v2 Set 97907_PM_at - -
          Mouse Genome 430A 2.0 Array 1417313_PM_at - -
          Mouse Genome 430A 2.0 Array 1417313_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 366364 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320332 - -
          Illumina mouse-6 v1.0 expression beadchip 2320332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233901 - -
          mouse-6 v1.1 (Illumina) scl066094.3_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066094.3_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233901 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233901 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066094.3_4-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0