Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01149632_g1
          See other PPAP2C GE Assays ›
          Gene Symbol:
          Ppap2c
          Gene Name:
          phosphatidic acid phosphatase type 2C
          Gene Aliases:
          Lpp2, PAP2-G, PAP2-gamma, PAP2c
          Chromosome Location:
          Chr.10: 79526424 - 79533787 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01149632_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppap2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005452.1 - 5 - 6 781 96
          AK049581.1 - 5 - 6 824 96
          AK185918.1 - 1 - 2 163 96
          BC010332.1 - 5 - 6 802 2654393 96
          BC016635.1 - 5 - 6 752 96
          BC085104.1 - 5 - 6 740 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002467.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277400 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277398 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277399 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277402 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277401 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129437 BB737982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129437 NM_015817.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631736 AK052443.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631736 DQ286956.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631736 NM_001103168.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631736 XM_001475156.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631736 XM_001475715.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680868 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114609 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129437 BB737982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129437 NM_015817.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P607674 BC085104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P607674 AK208995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P607674 BC010332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P607674 BC016635.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P607674 NM_015817.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27277410 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07119744 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114609 - -
          Whole Mouse Genome G4122A A_52_P607674 BC085104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607674 AK208995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607674 BC010332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607674 BC016635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607674 NM_015817.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107801 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134696 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114609 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07119744 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08838774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08838799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08838773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08838793 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08838777 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08838794 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277402 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277401 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277399 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277398 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27277400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08543825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08277860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08838769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08277859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08430398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08223190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08838783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08838786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08838773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08430410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08838777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08733139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08543820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08838799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08748493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08898047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08223202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08898055 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370553 - -
          Affymetrix HT MG-430 PM Array Plate 1451210_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420910_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370552 - -
          Mouse Genome 430 2.0 Array 1451210_at - -
          Mouse Genome 430 2.0 Array 1451210_PM_at - -
          Mouse Genome 430 2.0 Array 1420910_PM_at - -
          Mouse Genome 430 2.0 Array 1420910_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50784_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50784_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000017963.mm.1 TC1000002467.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017963.mm.1 NM_015817.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017961.mm.1 TC1000002467.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017961.mm.1 NM_015817.3 Refseq
          Mouse Expression Set 430 1451210_PM_at - -
          Mouse Expression Set 430 1420910_at - -
          Mouse Expression Set 430 1451210_at - -
          Mouse Expression Set 430 1420910_PM_at - -
          Mouse Genome 430A 2.0 Array 1451210_at - -
          Mouse Genome 430A 2.0 Array 1420910_PM_at - -
          Mouse Genome 430A 2.0 Array 1420910_at - -
          Mouse Genome 430A 2.0 Array 1451210_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451210_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451210_at - -
          Affymetrix HT Mouse Genome 430A Array 1420910_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420910_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107100711 - -
          Illumina mouse-6 v1.0 expression beadchip 2970348 - -
          Illumina mouse-6 v1.0 expression beadchip 107100711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655746 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249666 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655746 - -
          mouse-6 v1.1 (Illumina) scl37761.6_244-S - -
          mouse-6 v1.1 (Illumina) scl0050784.2_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 107100711 - -
          Illumina Sentrix mouse-6.v1 bead array 2970348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0050784.2_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970348 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107100711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3003631 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3003631 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249666 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216282 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-5x86c:80/100.66.135.11:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0